PA1-28256 detects DGCR6 from human samples.
PA1-28256 has been successfully used in Western blot applications.
The PA1-28256 immunogen is: Amino acids 1-220 of DGCR6.
DGCR6 may play a role in neural crest cell migration into the third and fourth pharyngeal pouches.