FIGURE: 1 / 3
Sequence of this protein is as follows: MNWELLLWLL VLCALLLLLV QLLRFLRADG DLTLLWAEWQ GRRPEWELTD MVVWVTGASS GIGEELAYQL SKLGVSLVLS ARRVHELERV KRRCLENGNL KEKDILVLPL DLTDTGSHEA ATKAVLQEFG RIDILVNNGG MSQRSLCMDT SLDVYRKLIE LNYLGTVSLT KCVLPHMIER KQGKIVTVNS ILGIISVPLS IGYCASKHAL RGFFNGLRTE LATYPGIIVS NICPGPVQSN IVENSLAGEV TKTIGNNGDQ SHKMTTSRCV RLMLISMAND LKEVWISEQP FLLVTYLWQY MPTWAWWITN KMGKKRIENF KSGVDADSSY FKIFKTKHD
Short-chain dehydrogenases/reductases (SDRs), such as DHRS7, catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids (Haeseleer and Palczewski, 2000 [PubMed 10800688]).[supplied by OMIM, Jun 2009].