Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DHX9 Antibodies

          Proteintech

          DHX9 Polyclonal Antibody

          40 References
          View all (21) DHX9 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite DHX9 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          DHX9 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DHX9 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DHX9 Antibody (17721-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells, using DHX9 antibody 17721-1-AP at 1:100 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DHX9 Antibody in Immunocytochemistry (ICC/IF)
          DHX9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DHX9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DHX9 Antibody in Western Blot (WB)
          DHX9 Antibody in Western Blot (WB)
          DHX9 Antibody in Western Blot (WB)
          DHX9 Antibody in Western Blot (WB)
          DHX9 Polyclonal Antibody

          Product Details

          17721-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 26 publications 26 publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 4 publications 4 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          -
          View 2 publications 2 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication

          RNA Immunoprecipitation (RIP)

          Assay-dependent
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DHX9 Fusion Protein Ag12104 (1-300 aa encoded by BC014246)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.21 mg/mL

          Amount

          31.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MGDVKNFLY AWCGKRKMTP SYEIRAVGNK NRQKFMCEVQ VEGYNYTGMG NSTNKKDAQS NAARDFVNYL VRINEIKSEE VPAFGVASPP PLTDTPDTTA NAEGDLPTTM GGPLPPHLAL KAENNSEVGA SGYGVPGPTW DRGANLKDYY SRKEEQEVQA TLESEEVDLN AGLHGNWTLE NAKARLNQYF QKEKIQGEYK YTQVGPDHNR SFIAEMTIYI KQLGRRIFAR EHGSNKKLAA QSCALSLVRQ LYHLGVVEAY SGLTKKKEGE TVEPYKVNLS QDLEHQLQNI IQELNLEILP P (1-300 aa encoded by BC014246)

          Target Information

          The human alanyl-tRNA synthetase (AARS) belongs to a family of tRNA synthases, of the class II enzymes. Class II tRNA synthases evolved early in evolution and are highly conserved. This is reflected by the fact that 498 of the 968-residue polypeptide human AARS shares 41% identity witht the E.coli protein. tRNA synthases are the enzymes that interpret the RNA code and attach specific aminoacids to the tRNAs that contain the cognate trinucleotide anticodons. They consist of a catalytic domain which interacts with the amino acid acceptor-T psi C helix of the tRNA, and a second domain which interacts with the rest of the tRNA structure.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ATP-dependent RNA helicase A; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9; DEAH (Asp-Glu-Ala-His) box helicase 9; DEAH (Asp-Glu-Ala-His) box polypeptide 9; DEAH box protein 9; DEAH-box helicase 9; DExH-box helicase 9; FLJ17406; Leukophysin; LKP; mHEL-5; NDH II; Nuclear DNA helicase II; OTTHUMP00000033287; RHA; RNA helicase; RNA helicase A; unnamed protein product

          View more View less

          Gene Aliases: AI326842; DDX9; DHX9; HEL-5; LKP; mHEL-5; MRD75; NDH2; NDHII; RHA

          View more View less

          UniProt ID: (Human) Q08211, (Mouse) O70133

          View more View less

          Entrez Gene ID: (Human) 1660, (Mouse) 13211, (Rat) 304859

          View more View less

          Function(s)
          nucleotide binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II core binding regulatory region RNA binding nucleic acid binding DNA binding DNA helicase activity DNA replication origin binding double-stranded DNA binding single-stranded DNA binding transcription cofactor activity transcription coactivator activity RNA binding RNA helicase activity double-stranded RNA binding single-stranded RNA binding mRNA binding helicase activity protein binding ATP binding hydrolase activity ATPase activity nucleoside-triphosphatase activity chromatin DNA binding 3'-5' DNA/RNA helicase activity 3'-5' RNA helicase activity siRNA binding RNA stem-loop binding ribosome binding 3'-5' DNA helicase activity triplex DNA binding metal ion binding nucleoside-triphosphate diphosphatase activity RNA polymerase II sequence-specific DNA binding transcription factor binding importin-alpha family protein binding RNA polymerase binding catalytic activity, acting on a nucleic acid RISC complex binding single-stranded DNA-dependent ATP-dependent 3'-5' DNA helicase activity sequence-specific mRNA binding promoter-specific chromatin binding RNA polymerase II transcription factor binding ATP-dependent RNA helicase activity ATP-dependent helicase activity poly(A) RNA binding molecular_function RNA helicase
          Process(es)
          alternative mRNA splicing, via spliceosome osteoblast differentiation immune system process DNA replication chromatin organization DNA-templated transcription, termination regulation of transcription from RNA polymerase II promoter mRNA processing regulation of translation inflammatory response RNA splicing gene silencing by RNA positive regulation of interferon-alpha production positive regulation of interferon-beta production positive regulation of interleukin-18 production positive regulation of interleukin-6 production positive regulation of tumor necrosis factor production gene silencing by miRNA DNA-templated viral transcription innate immune response positive regulation of innate immune response positive regulation of DNA repair positive regulation of DNA replication positive regulation of transcription from RNA polymerase II promoter positive regulation of RNA export from nucleus positive regulation of fibroblast proliferation rhythmic process positive regulation of viral transcription regulation of mRNA processing regulation of defense response to virus by host positive regulation of inflammatory response mRNA transport positive regulation of NF-kappaB transcription factor activity positive regulation of response to cytokine stimulus protein-containing complex assembly pyroptosis small RNA loading onto RISC CRD-mediated mRNA stabilization cellular response to exogenous dsRNA negative regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay protein localization to cytoplasmic stress granule regulation of cytoplasmic translation positive regulation of cytoplasmic translation RNA processing circadian rhythm cellular response to heat biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline