Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SMAC Antibodies

          Proteintech

          DIABLO Polyclonal Antibody

          11 References
          View all (29) SMAC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite DIABLO Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (29)
          DIABLO Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DIABLO Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 29

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DIABLO Antibody (10434-1-AP) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using DIABLO antibody (Product # 10434-1-AP) at dilution of 1:200 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DIABLO Antibody in Immunocytochemistry (ICC/IF)
          DIABLO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIABLO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIABLO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIABLO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIABLO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIABLO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIABLO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIABLO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIABLO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIABLO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIABLO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIABLO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIABLO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIABLO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DIABLO Antibody in Western Blot (WB)
          DIABLO Antibody in Western Blot (WB)
          DIABLO Antibody in Western Blot (WB)
          DIABLO Antibody in Western Blot (WB)
          DIABLO Antibody in Western Blot (WB)
          DIABLO Antibody in Western Blot (WB)
          DIABLO Antibody in Western Blot (WB)
          DIABLO Antibody in Western Blot (WB)
          DIABLO Antibody in Western Blot (WB)
          DIABLO Antibody in Western Blot (WB)
          DIABLO Antibody in Western Blot (WB)
          DIABLO Antibody in Flow Cytometry (Flow)
          DIABLO Antibody in Immunoprecipitation (IP)
          DIABLO Antibody in Immunoprecipitation (IP)
          DIABLO Polyclonal Antibody

          Product Details

          10434-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 10 publications 10 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:200
          -

          Flow Cytometry (Flow)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DIABLO Fusion Protein Ag0696 (1-239 aa encoded by BC011909)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Amount

          40.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody recognizes all the three isoforms (27 kDa and 22 kDa) of DIABLO. The highest expression was in the testis, adrenal gland, and ears.

          Immunogen sequence: MAALKSWLS RSVTSFFRYR QCLCVPVVAN FKKRCFSELI RPWHKTVTIG FGVTLCAVPI AQKSEPHSLS SEALMRRAVS LVTDSTSTFL SQTTYALIEA ITEYTKAVYT LTSLYRQYTS LLGKMNSEEE DEVWQVIIGA RAEMTSKHQE YLKLETTWMT AVGLSEMAAE AAYQTGADQA SITARNHIQL VKLQVEEVHQ LSRKAETKLA EAQIEELRQK TQEEGEERAE SEQEAYLRED (1-239 aa encoded by BC011909)

          Target Information

          DIABLO (smac), a mitochondrial protein, activates various forms of apoptosis. This activation may be due to the neutralization of one or more members of the IAP family of apoptosis inhibitory proteins. Smac exits the mitochondria and enter the cytosol during certain apoptosis triggered events. Mitochondrial-mediated apoptosis is important in animal development and tissue homeostasis, with alterations resulting in a range of malignant disorders. Upon apoptotic stimuli, the mitochondrial proteins cytochrome c and Smac/DIABLO are released into the cytosol. The release of these proteins, however, occurs via different mechanisms. Smac/DIABLO has also been found to play a key role in regulating the sensitization of cancer cells to apoptosis (both immune and drug-induced).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: antagonist of IAPs (inhibitors of apoptosis); diablo homolog (Drosophila) isoform DIABLO_1; diablo homolog (Drosophila) isoform DIABLO_2; Diablo homolog, mitochondrial; Diablo IAP-binding mitochondrial protein; diablo-like protein; DIABLO-S; direct IAP binding protein with low PI; Direct IAP-binding protein with low pI; FLJ10537; FLJ25049; hCG_1782202; Second mitochondria-derived activator of caspase; Second mitochondria-derived activator of caspases; short isoform; SMAC; Smac protein; SMAC3; unnamed protein product

          View more View less

          Gene Aliases: 0610041G12Rik; 1700006L01Rik; AU040403; DFNA64; DIABLO; SMAC

          View more View less

          UniProt ID: (Human) Q9NR28, (Mouse) Q9JIQ3

          View more View less

          Entrez Gene ID: (Human) 56616, (Mouse) 66593, (Rat) 288753

          View more View less

          Function(s)
          protein binding
          Process(es)
          apoptotic process activation of cysteine-type endopeptidase activity involved in apoptotic process intrinsic apoptotic signaling pathway in response to oxidative stress activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c positive regulation of apoptotic process neuron apoptotic process intrinsic apoptotic signaling pathway extrinsic apoptotic signaling pathway via death domain receptors apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline