Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • DIAPH3 Antibodies

          Abnova

          DIAPH3 Polyclonal Antibody, MaxPab™

          View all (13) DIAPH3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite DIAPH3 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          DIAPH3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DIAPH3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DIAPH3 Antibody (H00081624-D01P) in WB

          Western Blot analysis of DIAPH3 expression in transfected 293T cell line by DIAPH3 MaxPab polyclonal antibody. Lane 1: DIAPH3 transfected lysate(98.10 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DIAPH3 Antibody in Western Blot (WB)
          DIAPH3 Polyclonal Antibody, MaxPab™

          Product Details

          H00081624-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DIAPH3 (NP_112194.2, 1 a.a. approximately 849 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSEERSLSLL AKAVDPRHPN MMTDVVKLLS AVCIVGEESI LEEVLEALTS AGEEKKIDRF FCIVEGLRHN SVQLQVACMQ LINALVTSPD DLDFRLHIRN EFMRCGLKEI LPNLKCIKND GLDIQLKVFD EHKEEDLFEL SHRLEDIRAE LDEAYDVYNM VWSTVKETRA EGYFISILQH LLLIRNDYFI RQQYFKLIDE CVSQIVLHRD GMDPDFTYRK RLDLDLTQFV DICIDQAKLE EFEEKASELY KKFEKEFTDH QETQAELQKK EAKINELQAE LQAFKSQFGA LPADCNIPLP PSKEGGTGHS ALPPPPPLPS GGGVPPPPPP PPPPPLPGMR MPFSGPVPPP PPLGFLGGQN SPPLPILPFG LKPKKEFKPE ISMRRLNWLK IRPHEMTENC FWIKVNENKY ENVDLLCKLE NTFCCQQKER REEEDIEEKK SIKKKIKELK FLDSKIAQNL SIFLSSFRVP YEEIRMMILE VDETRLAESM IQNLIKHLPD QEQLNSLSQF KSEYSNLCEP EQFVVVMSNV KRLRPRLSAI LFKLQFEEQV NNIKPDIMAV STACEEIKKS KSFSKLLELV LLMGNYMNAG SRNAQTFGFN LSSLCKLKDT KSADQKTTLL HFLVEICEEK YPDILNFVDD LEPLDKASKV SVETLEKNLR QMGRQLQQLE KELETFPPPE DLHDKFVTKM SRFVISAKEQ YETLSKLHEN MEKLYQSIIG YYAIDVKKVS VEDFLTDLNN FRTTFMQAIK ENIKKREAEE KEKRVRIAKE LAERERLERQ QKKKRLLEMK TEGDETGVMD NLLEALQSGA AFRDRRKRTP MPKDVRQSLS PMSQRPVLKV CNHGNKPYL

          Target Information

          This gene encodes a member of the diaphanous subfamily of the formin family. Members of this family are involved in actin remodeling and regulate cell movement and adhesion. Mutations in this gene are associated with autosomal dominant auditory neuropathy 1. Multiple transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: actin binding protein; Drf3; hDia3; diaphanous homolog 3; Diaphanous-related formin-3; DKFZp434C0931; DKFZp686A13178; DRF3; FLJ34705; MDia2; Protein diaphanous homolog 3; unnamed protein product

          View more View less

          Gene Aliases: AN; AUNA1; DIA2; DIAP3; DIAPH3; DRF3; mDia2; NSDAN

          View more View less

          UniProt ID: (Human) Q9NSV4

          View more View less

          Entrez Gene ID: (Human) 81624

          View more View less

          Function(s)
          actin binding microtubule binding cytoskeletal protein binding small GTPase binding protein homodimerization activity cadherin binding
          Process(es)
          microtubule cytoskeleton organization in utero embryonic development cytoskeleton organization actin filament organization negative regulation of microtubule depolymerization chromosome segregation integrin-mediated signaling pathway sensory perception of sound gene expression endosomal transport establishment of cell polarity cell projection organization actin cytoskeleton organization actin filament polymerization neuron differentiation erythrocyte differentiation macrophage differentiation protein-containing complex remodeling erythrocyte enucleation actin nucleation microtubule polymerization actin filament bundle assembly actin crosslink formation inner ear receptor cell differentiation head development autophagosome-lysosome fusion podosome assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline