PA1-27490 detects DLG7 from human samples.
PA1-27490 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.
The PA1-27556 immunogen is a synthetic peptide corresponding to human Discs large homolog 7.
Store at 4°C short term; for long term storage, store at -20°C, avoiding freeze/thaw cycles.
DLG7 is a potential cell cycle regulator that may play a role in carcinogenesis of cancer cells and mitotic phosphoprotein regulated by the ubiquitin-proteasome pathway. The protein is also a key regulator of adherens junction integrity and differentiation that may be involved in CDH1-mediated adhesion and signaling in epithelial cells.