Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • DLL1 Antibodies

          Invitrogen

          DLL1 Polyclonal Antibody

          View all (22) DLL1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite DLL1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          DLL1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DLL1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DLL1 Antibody (PA5-85819) in WB

          Western blot analysis of DLL1 in mouse tissue extract using DLL1 polyclonal antibody (Product # PA5-85819) using 50 µg of sample at a dilution of 1:100. Sample was then incubated with HRP-conjugated anti-rabbit IgG secondary antibody. Prior to incubation with primary antibody, the sample was separated on 7.5% SDS-PAGE. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DLL1 Antibody in Western Blot (WB)

          Product Details

          PA5-85819

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein encompassing a sequence within the Extracellular domain of human DLL1. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.81 mg/mL

          Amount

          81 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 20% glycerol

          Contains

          0.025% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2792955

          Product Specific Information

          Keep as concentrated solution.

          Predicted reactivity: Mouse (87%), Rat (84%), Chicken (81%).

          Positive Control: mouse heart.

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          Delta-like protein 1 (DLL1) is a transmembrane protein characterized by epidermal growth factor (EGF)-like repeats. It is a ligand for Notch receptor. It blocks the differentiation of progenitor cells into the B-cell lineage while promoting the emergence of a population of cells with the characteristics of a T-cell/NK-cell precursor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: delta like-1; Delta-like protein 1; Delta1; Drosophila Delta homolog 1; epididymis secretory sperm binding protein; H-Delta-1; Notch ligand; OTTHUMP00000017690; unnamed protein product

          View more View less

          Gene Aliases: Delta; DELTA1; DL1; DLL1; NEDBAS; UNQ146/PRO172

          View more View less

          UniProt ID: (Human) O00548, (Mouse) Q61483

          View more View less

          Entrez Gene ID: (Human) 28514, (Mouse) 13388

          View more View less

          Function(s)
          Notch binding calcium ion binding protein binding Tat protein binding receptor agonist activity scaffold protein binding
          Process(es)
          in utero embryonic development somitogenesis somite specification heart looping cell communication Notch signaling pathway cell-cell signaling multicellular organism development determination of left/right symmetry compartment pattern specification nervous system development organ morphogenesis auditory receptor cell fate commitment cell differentiation negative regulation of interleukin-10 production inner ear morphogenesis odontogenesis of dentin-containing tooth auditory receptor cell differentiation negative regulation of cell differentiation negative regulation of auditory receptor cell differentiation negative regulation of myeloid cell differentiation negative regulation of neuron differentiation positive regulation of Notch signaling pathway positive regulation of transcription from RNA polymerase II promoter inner ear development regulation of neurogenesis Notch signaling involved in heart development left/right axis specification cell fate determination inhibition of neuroepithelial cell differentiation marginal zone B cell differentiation type B pancreatic cell development regulation of blood pressure positive regulation of cell proliferation negative regulation of cell proliferation proximal/distal pattern formation positive regulation of gene expression astrocyte development regulation of somitogenesis spinal cord development cerebellar molecular layer formation cerebellar Purkinje cell layer structural organization hemopoiesis myeloid cell differentiation regulation of cell adhesion neuron differentiation negative regulation of epithelial cell differentiation regulation of vascular endothelial growth factor receptor signaling pathway negative regulation of glial cell apoptotic process organ growth regulation of growth negative regulation of epidermal cell differentiation negative regulation of myoblast differentiation negative regulation of Notch signaling pathway positive regulation of endocytosis lateral inhibition skeletal muscle tissue growth regulation of skeletal muscle tissue growth positive regulation of skeletal muscle tissue growth neuron fate specification regulation of cell division retina development in camera-type eye retina morphogenesis in camera-type eye neuroepithelial cell differentiation Notch signaling pathway involved in arterial endothelial cell fate commitment nephron development proximal tubule development loop of Henle development clathrin-mediated endocytosis energy homeostasis endothelial tip cell fate specification neuronal stem cell population maintenance skin epidermis development regulation of vascular endothelial growth factor signaling pathway positive regulation of sprouting angiogenesis negative regulation of cardiac muscle cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline