Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HSP40 Antibodies

          Proteintech

          DNAJB1 Polyclonal Antibody, CoraLite® Plus 647

          View all (51) HSP40 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite DNAJB1 Polyclonal Antibody, CoraLite® Plus 647

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          DNAJB1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DNAJB1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DNAJB1 Antibody (CL647-13174) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using CoraLite® Plus 647 DNAJB1 antibody (CL647-13174) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DNAJB1 Antibody in Immunocytochemistry (ICC/IF)
          DNAJB1 Antibody in Flow Cytometry (Flow)
          DNAJB1 Polyclonal Antibody, CoraLite® Plus 647

          Product Details

          CL647-13174

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cell suspension
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein
          3D Epitope / Immunogen

          Conjugate

          CoraLite® Plus 647 CoraLite® Plus 647 CoraLite® Plus 647

          Excitation/Emission Max

          654/674 nm

          Form

          Liquid

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 50% glycerol, 0.5% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MGKDYYQTLG LARGASDEEI KRAYRRQALR YHPDKNKEPG AEEKFKEIAE AYDVLSDPRK REIFDRYGEE GLKGSGPSGG SGGGANGTSF SYTFHGDPHA MFAEFFGGRN PFDTFFGQRN GEEGMDIDDP FSGFPMGMGG FTNVNFGRSR SAQEPARKKQ DPPVTHDLRV SLEEIYSGCT KKMKISHKRL NPDGKSIRNE DKILTIEVKK GWKEGTKITF PKEGDQTSNN IPADIVFVLK DKPHNIFKRD GSDVIYPARI SLREALCGCT VNVPTLDGRT IPVVFKDVIR PGMRRKVPGE GLPLPKTPEK RGDLIIEFEV IFPERIPQTS RTVLEQVLPI

          Target Information

          TCF-9 (transcription factor 9), also known as C2orf3 (chromosome 2 open reading frame 3), GCF (GC-rich sequence DNA-binding factor) or DNABF, is a 781 amino acid nuclear protein that belongs to the GCF family. Widely expressed, GCF binds the GC-rich sequences of beta-Actin, EGFR and calcium-dependent protease (CANP) promoters. GCF contains multiple phosphoserine and phosphothreonine residues, and two GCF isoforms are produced due to alternative splicing events. GCF is considered a candidate for susceptibility to dyslexia (DYX3) as both genes reside in close proximity on human chromosome 2. Chromosome 2 is the second largest human chromosome and consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DnaJ (Hsp40) homolog, subfamily B, member 1; DnaJ homolog subfamily B member 1; DnaJ protein homolog 1; DNAJB1; hDj-1; Heat shock 40 kDa protein 1; Heat shock protein; HSP; HSP 40; HSP40; Human DnaJ protein 1; NDAJB1; radial spoke 16 homolog B; unnamed protein product

          View more View less

          Gene Aliases: 0610007I11Rik; DjB1; DNAJ1; DNAJB1; HDJ1; Hsp40; HSPF1; RSPH16B; Sis1

          View more View less

          UniProt ID: (Human) P25685, (Mouse) Q9QYJ3

          View more View less

          Entrez Gene ID: (Human) 3337, (Rat) 361384, (Mouse) 81489

          View more View less

          Function(s)
          ATPase activator activity transcription corepressor activity protein binding Hsp70 protein binding protein binding involved in protein folding cadherin binding unfolded protein binding chaperone binding ATPase binding transcription regulator inhibitor activity chaperone
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter protein folding response to unfolded protein forebrain development cellular response to heat negative regulation of inclusion body assembly regulation of cellular response to heat positive regulation of protein folding positive regulation of ATPase activity chaperone mediated protein folding requiring cofactor chaperone cofactor-dependent protein refolding
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline