Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: GRDFYKILG VPRSASIKDI KKAYRKLALQ LHPDRNPDDP QAQEKFQDLG AAYEVLSDSE KRKQYDTYGE EGLKDGHQSS HGDIFSHFFG DFGFMFGGTP RQQDRNIPRG SDIIVDLEVT LEEVYAGNFV EVVRNKPVAR QAPGKRKCNC RQEMRTTQLG PGRFQMTQEV VCDECPNVKL VNEERTLEVE IEPGVRDGME YPFIGEGEPH VDGEPGDLRF RIKVVKHPIF ERRGDDLYTN VTISLVESLV GFEMDITHLD GHKVHISRDK ITRPGAKLWK KGEGLPNFDN NNIKGSLIIT FDVDFPKEQL TEEAREGIKQ LLKQGSVQKV YNGLQGY (23-358 aa encoded by BC001144)
DNAJB11 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity bystimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus a glycine/phenylalanine (G/F)-rich region and a C-terminal cysteine-rich region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APOBEC1-binding protein 2; DnaJ (Hsp40) homolog, subfamily B, member 11; DnaJ protein; DnaJ protein 9; DNAJB11; endoplasmic reticulum DNA J domain-containing protein 3; ER-associated DNAJ protein 3; ER-associated Hsp40 co-chaperone; ER-resident protein ERdj3; ERdj3; ERj3p; HEDJ; liver regeneration-related protein LRRGT00084; PWP1-interacting protein 4; unnamed protein product
Gene Aliases: 1810031F23Rik; ABBP-2; ABBP2; Dj-9; DJ9; EDJ; ERdj3; ERj3; ERj3p; LRRGT00084; PKD6; PRO1080; UNQ537
UniProt ID: (Human) Q9UBS4, (Rat) Q6TUG0, (Mouse) Q99KV1
Entrez Gene ID: (Human) 51726, (Rat) 360734, (Mouse) 67838
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support