Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • DRD1 Antibodies

          Invitrogen

          DRD1 Polyclonal Antibody

          1 Reference
          View all (22) DRD1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite DRD1 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          DRD1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DRD1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DRD1 Antibody (720276) in WB

          Western blot analysis was performed on whole cell extracts (30 µg lysate) of U-2 OS (Lane 1), Tissue lysates of (30 µg lysate) Mouse Stomach (Lane 2), Rat Stomach (Lane 3), Mouse Brain (Lane 4). The blots were probed with Anti-Dopamine receptor D1 Polyclonal Antibody (Product # 720276, 1-2 µg/mL) and detected by chemiluminescence using Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, HRP conjugate (Product # A27036, 0.4 µg/mL, 1:2500 dilution). A 48 kDa band corresponding to Dopamine receptor D1 was observed. Known quant... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DRD1 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          DRD1 Polyclonal Antibody

          Product Details

          720276

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-2 µg/mL
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide corresponding to Human DRD1 (aa 164-179)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2-7.4, with 0.1% BSA, 30% glycerol

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2633231

          Product Specific Information

          These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature.

          This antibody is predicted to react with Monkey, Cat, Bovine and Pig.

          Target Information

          The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: D1 receptor; D1A dopamine receptor; D1a receptor; Dopamine 1 receptor; dopamine D1 receptor; dopamine receptor D1A; Dopamine-1A receptor; DRD1; G protein-coupled receptor DRD1; G-protein coupled receptor; unnamed protein product

          View more View less

          Gene Aliases: C030036C15Rik; D1a; D1R; DADR; Drd-1; DRD1A; Gpcr15

          View more View less

          UniProt ID: (Human) P21728, (Mouse) Q61616

          View more View less

          Entrez Gene ID: (Human) 1812, (Rat) 24316, (Mouse) 13488

          View more View less

          Function(s)
          dopamine neurotransmitter receptor activity, coupled via Gs G-protein alpha-subunit binding G-protein coupled receptor activity dopamine neurotransmitter receptor activity protein binding heterotrimeric G-protein binding dopamine binding arrestin family protein binding G-protein coupled serotonin receptor activity receptor binding protein phosphatase binding neurotransmitter receptor activity angiotensin receptor binding D3 dopamine receptor binding macromolecular complex binding ATPase binding heterocyclic compound binding
          Process(es)
          temperature homeostasis conditioned taste aversion behavioral fear response regulation of protein phosphorylation positive regulation of protein phosphorylation synaptic transmission, dopaminergic response to amphetamine protein phosphorylation negative regulation of protein kinase activity protein import into nucleus intracellular protein transport muscle contraction G-protein coupled receptor signaling pathway G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger adenylate cyclase-modulating G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway activation of adenylate cyclase activity adenylate cyclase-activating dopamine receptor signaling pathway dopamine receptor signaling pathway chemical synaptic transmission synapse assembly learning memory mating behavior grooming behavior locomotory behavior adult walking behavior feeding behavior associative learning visual learning response to xenobiotic stimulus astrocyte development response to activity transmission of nerve impulse neuronal action potential regulation of vasoconstriction calcium-mediated signaling dentate gyrus development striatum development hippocampus development orbitofrontal cortex development cerebral cortex GABAergic interneuron migration positive regulation of cell migration negative regulation of cell migration peristalsis response to food response to estradiol response to retinoic acid cellular response to insulin stimulus response to nicotine operant conditioning social behavior regulation of locomotion regulation of dopamine metabolic process response to cocaine vasodilation negative regulation of circadian sleep/wake cycle, sleep maternal behavior eating behavior response to amino acid regulation of ion transport response to ethanol positive regulation of membrane potential habituation sensitization behavioral response to cocaine regulation of synaptic plasticity response to steroid hormone negative regulation of synaptic transmission positive regulation of release of sequestered calcium ion into cytosol positive regulation of synaptic transmission, glutamatergic phospholipase C-activating dopamine receptor signaling pathway long-term synaptic potentiation long term synaptic depression cellular response to hypoxia cellular response to catecholamine stimulus G protein-coupled serotonin receptor signaling pathway glucose import across plasma membrane modification of postsynaptic structure postsynaptic modulation of chemical synaptic transmission presynaptic modulation of chemical synaptic transmission negative regulation of cellular response to hypoxia positive regulation of long-term synaptic potentiation positive regulation of feeding behavior phospholipase C-activating G-protein coupled receptor signaling pathway dopamine metabolic process positive regulation of potassium ion transport positive regulation of MAPK cascade regulation of dopamine uptake involved in synaptic transmission adenylate cyclase-activating adrenergic receptor signaling pathway negative regulation of behavioral fear response
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch