PA5-19429 targets Dicer in WB applications and shows reactivity with Drosophila samples.
The PA5-19429 immunogen is synthetic peptide derived from residues 900 - 1000 Dicer (Dcr1).
PA5-19429 detects Dicer which has a predicted molecular weight of approximately 255 kDa.
This gene encodes a protein possessing an RNA helicase motif containing a DEXH box in its amino terminus and an RNA motif in the carboxy terminus. The encoded protein functions as a ribonuclease and is required by the RNA interference and small temporal RNA pathways to produce the active small RNA component that represses gene expression. Alternative splicing results in multiple transcript variants.