Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Dinitrophenyl-KLH Antibodies

          Invitrogen

          Dinitrophenyl-KLH Polyclonal Antibody, Alexa Fluor™ 488

          4 Published Figures
          31 References
          View all (3) Dinitrophenyl-KLH antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Dinitrophenyl-KLH Polyclonal Antibody, Alexa Fluor™ 488

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Published Figures (4)
          Dinitrophenyl-KLH Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Dinitrophenyl-KLH Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Dinitrophenyl-KLH Antibody (A-11097) in ICC/IF

          American journal of physiology. Cell physiology 2011 - Published figure using Dinitrophenyl-KLH polyclonal antibody (Product # A-11097) in Immunocytochemistry {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Dinitrophenyl-KLH Antibody in Immunocytochemistry (ICC/IF)
          Dinitrophenyl-KLH Antibody in Immunohistochemistry (IHC)
          Dinitrophenyl-KLH Antibody in Flow Cytometry (Flow)
          Dinitrophenyl-KLH Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          A-11097

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 3 publications 3 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 14 publications 14 publications

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 7 publications 7 publications

          Flow Cytometry (Flow)

          -
          View 5 publications 5 publications

          In Situ Hybridization (ISH)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Chemical

          Published species

          Chemical

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Dinitrophenyl-KLH

          Conjugate

          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488

          Excitation/Emission Max

          499/520 nm View spectra spectra

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          1 mg

          Purification

          purified

          Storage buffer

          PBS, pH 7.5

          Contains

          5mM sodium azide

          Storage conditions

          4°C, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2534131

          Product Specific Information

          The anti-DNP-KLH antibody is supplied in 0.1 M sodium phosphate, 0.1 M sodium chloride, pH 7.5, containing 5 mM sodium azide. The degree of labeling is indicated on the product label. This antibody was raised in rabbit against DNP conjugated to keyhole limpet hemocyanin. The IgG fraction, which contains both anti-DNP and anti-KLH IgGs, was isolated from total serum and determined to be >95% pure by SDS-polyacrylamide gel electrophoresis. It is a good practice to centrifuge protein conjugate solutions briefly in a microcentrifuge before use; only the supernatant should then be added to the experiment. This step will eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically. Peak absorption and fluorescence emission wavelengths for Alexa Fluor™ 488 anti-DNPKLH conjugate is 494 nm and 520 nm, respectively

          Because of their high affinity for the dinitrophenyl hapten1,2 (DNP), Invitrogen's antibodies against the dinitrophenyl- keyhole limpet hemocyanin conjugate (anti-DNP-KLH) are sensitive probes for detecting DNP-labeled molecules. The fluorescent and biotinylated anti-DNP-KLH conjugates can serve as second-step reagents for signal amplification in protein and nucleic acid detection systems, including in situ hybridization studies employing DNP-modified DNA probes. Because anti-DNP antibody binds well to DAMP, an acidotropic reagent that can penetrate cell membranes, anti-DNP-KLH antibodies can also be used with DAMP for immunochemical localization of intracellular compartments with low pH. In addition, anti-DNP-KLH antibodies may recognize compounds with a trinitrophenyl group, allowing the isolation of cell-surface molecules of human melanoma cells that have been labeled with trinitrobenzene sulfonic acid (TNBS).

          Target Information

          DNP-KLH antibodies are used for detecting DNP-labeled molecules. Fluorescent and biotinylated anti-DNP-KLH conjugates can serve as second-step reagents for signal amplification in protein and nucleic acid detection systems, including in situ hybridization studies employing DNP-modified DNA probes. Because anti-DNP antibody binds well to DAMP (D-1552), an acidotropic reagent that can penetrate cell membranes, anti-DNP-KLH antibodies can also be used with DAMP for immunochemical localization of intracellular compartments with low pH. DNP-KLH antibodies may recognize compounds with a trinitrophenyl group, allowing the isolation of cell-surface molecules of human melanoma cells that have been labeled with trinitrobenzene sulfonic acid (TNBS).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: C6H3N2O4; DNP-KLH

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch