Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • EEF1A1 Antibodies

          Proteintech

          EEF1A1 Polyclonal Antibody

          25 References
          View all (24) EEF1A1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite EEF1A1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (22)
          EEF1A1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          EEF1A1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 22

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EEF1A1 Antibody (11402-1-AP) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using EEF1A1 antibody (Product # 11402-1-AP) at dilution of 1:200 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EEF1A1 Antibody in Immunocytochemistry (ICC/IF)
          EEF1A1 Antibody in Immunocytochemistry (ICC/IF)
          EEF1A1 Antibody in Immunocytochemistry (ICC/IF)
          EEF1A1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EEF1A1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EEF1A1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Antibody in Western Blot (WB)
          EEF1A1 Polyclonal Antibody

          Product Details

          11402-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 15 publications 15 publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 3 publications 3 publications

          Immunoprecipitation (IP)

          -
          View 2 publications 2 publications

          RNA Immunoprecipitation (RIP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          EEF1A1 Fusion Protein Ag1938 (144-462 aa encoded by BC014224)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.32 mg/mL

          Amount

          48 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human EEF1A1.

          Immunogen sequence: GVKQLIVGV NKMDSTEPPY SQKRYEEIVK EVSTYIKKIG YNPDTVAFVP ISGWNGDNML EPSANMPWFK GWKVTRKDGN ASGTTLLEAL DCILPPTRPT DKPLRLPLQD VYKIGGIGTV PVGRVETGVL KPGMVVTFAP VNVTTEVKSV EMHHEALSEA LPGDNVGFNV KNVSVKDVRR GNVAGDSKND PPMEAAGFTA QVIILNHPGQ ISAGYAPVLD CHTAHIACKF AELKEKIDRR SGKKLEDGPK FLKSGDAAIV DMVPGKPMCV ESFSDYPPLG RFAVRDMRQT VAVGVIKAVD KKAAGAGKVT KSAQKAQKAK (144-462 aa encoded by BC014224)

          Target Information

          This protein promotes the GTP-dependent binding of aminoacyl-tRNA to the A-site of ribosomes during protein biosynthesis. With PARP1 and TXK, forms a complex that acts as a T helper 1 (Th1) cell-specific transcription factor and binds the promoter of IFN-gamma to directly regulate its transcription, and is thus involved importantly in Th1 cytokine production.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cervical cancer suppressor 3; CTCL tumor antigen; eEF1A-1; EF-1 alpha2; EF-1-alpha-1; EF-Tu; EF1a-like protein; EF1A1; elongation factor 1 A-1; elongation factor 1 alpha; elongation factor 1 alpha subunit; elongation factor 1-alpha; Elongation factor 1-alpha 1; Elongation factor Tu; epididymis secretory sperm binding protein; Eukaryotic elongation factor 1 A-1; eukaryotic translation elongation factor 1 alpha 1-like 14; eukaryotic translation elongation factor 1 alpha 2 like 1; glucocorticoid receptor AF-1 specific elongation factor; GRAF 1EF; GRAF-1EF; leukocyte receptor cluster (LRC) member 7; Leukocyte receptor cluster member 7; prostate tumor-inducing protein 1; prostate tumor-inducing protein 1; PTI-1; translation elongation factor 1 alpha 1-like 14; unnamed protein product

          View more View less

          Gene Aliases: CCS-3; CCS3; EE1A1; EEF-1; EEF1A; eEF1A-1; EEF1A1; Eef1a2; Eef1a2l1; EF-Tu; EF1A; EF1A1; EF1alpha1; GRAF-1EF; LENG7; PTI1; SI

          View more View less

          UniProt ID: (Human) P68104, (Mouse) P10126, (Rat) P62630

          View more View less

          Entrez Gene ID: (Human) 1915, (Mouse) 13627, (Rat) 171361

          View more View less

          Function(s)
          tRNA binding nucleotide binding RNA binding translation elongation factor activity GTPase activity protein binding GTP binding hydrolase activity kinase activator activity kinase binding protein kinase binding binding, bridging mRNA binding poly(A) RNA binding translation factor
          Process(es)
          translation translational elongation positive regulation by host of viral genome replication cellular response to epidermal growth factor stimulus regulation of chaperone-mediated autophagy protein homooligomerization positive regulation of apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline