Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody is a rabbit polyclonal antibody raised against a region of human EGR2.
Immunogen sequence: LGVCTMSQT QPDLDHLYSP PPPPPPYSGC AGDLYQDPSA FLSAATTSTS SSLAYPPPPS YPSPKPATDP GLFPMIPDYP GFFPSQCQRD LHGTAGPDRK PFPCPLDTLR VPPPLTPLST IRKPFPCPLD TLRVPPPLTP LSTIRNFTLG GPSAGVTGPG ASGGSEGPRL PGSSSAAAAA AAAAAYNPHH LPLRPILRPR KYPNRPSKTP VHERPYPCPA EGCDRRFSRS DELTRHIRIH TGHKPFQCRI CMRNFSRSDH LTTHIRTHTG EKPFACDYCG RKFARSDERK RHTKIHLRQK ERKSSAPSAS VPAPSTASCS GGVQPGGTLC SSNSSSLGGG PLAPCSSRTR TP (149-499 aa encoded by BC035625)
The EGR2 gene provides instructions for making a protein called early growth response 2, which is part of the early growth response family of proteins. These proteins bind to specific areas of DNA and help control the activity of particular genes. On the basis of this action, the proteins are referred to as transcription factors. The early growth response 2 protein activates several genes that are involved in the formation and maintenance of myelin, the fatty substance that covers and protects nerve cells. Myelin promotes the efficient transmission of nerve impulses. If myelin is lost (demyelination) or its structure is disrupted, the transmission of nerve impulses is impaired. Mutations in the EGR2 gene can cause two forms of Charcot-Marie-Tooth disease, type 1D or type 4E (sometimes called congenital hypomyelinating neuropathy) or a severe form of type 1D (sometimes called Dejerine-Sottas syndrome) that begins during infancy or early childhood.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686J1957; E3 SUMO-protein transferase ERG2; early growth response 2 protein; early growth response protein 2; EGR-2; FLJ1454; FLJ14547; Krox-20 homolog; KROX-20, Drosophila, homolog (early growth response-2); OTTHUMP00000062133; unnamed protein product; zinc finger protein Krox-20
Gene Aliases: AT591; CMT1D; CMT4E; Egr-2; Krox-20; KROX20; NGF1-B; Zfp-25; Zfp-6
UniProt ID: (Human) P11161, (Rat) P51774, (Mouse) P08152
Entrez Gene ID: (Human) 1959, (Rat) 114090, (Mouse) 13654
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support