Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human EIF2A, recognizes specifically the 65 kDa EIF2A protein.
Immunogen sequence: IASTDVDKT GASYYGEQTL HYIATNGESA VVQLPKNGPI YDVVWNSSST EFCAVYGFMP AKATIFNLKC DPVFDFGTGP RNAAYYSPHG HILVLAGFGN LRGQMEVWDV KNYKLISKPV ASDSTYFAWC PDGEHILTAT CAPRLRVNNG YKIWHYTGSI LHKYDVPSNA ELWQVSWQPF LDGIFPAKTI TYQAVPSEVP NEEPKVATAY RPPALRNKPI TNSKLHEEEP PQNMKPQSGN DKPLSKTALK NQRKHEAKKA AKQEARSDKS PDLAPTPAPQ STPRNTVSQS ISGDPEIDKK IKNLKKKLKA IEQLKEQAAT GKQLEKNQLE KIQKETALLQ ELEDLKLGI (238-585 aa encoded by BC011885)
EIF2A is a 65-kD protein that catalyzes the formation of puromycin-sensitive 80S preinitiation complexes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 65 kDa eukaryotic translation initiation factor 2A; CDA02; EIF-2A; eIF2A; Eukaryotic translation initiation factor 2A; eukaryotic translation initiation factor 2A, 65kDa; MST089; MSTP004; MSTP089; unnamed protein product
Gene Aliases: CDA02; D030048D22; D3Ertd194e; EIF-2A; EIF2A; MST089; MSTP004; MSTP089
UniProt ID: (Human) Q9BY44, (Mouse) Q8BJW6
Entrez Gene ID: (Human) 83939, (Rat) 502531, (Mouse) 229317
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support