Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PKR Antibodies

          Proteintech

          EIF2AK2/PKR Polyclonal Antibody

          30 References
          View all (54) PKR antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite EIF2AK2/PKR Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (29)
          EIF2AK2/PKR Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          EIF2AK2/PKR Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 29

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EIF2AK2/PKR Antibody (18244-1-AP) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using EIF2AK2, PKR antibody (Product # 18244-1-AP) at dilution of 1:200 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EIF2AK2/PKR Antibody in Immunocytochemistry (ICC/IF)
          EIF2AK2/PKR Antibody in Immunocytochemistry (ICC/IF)
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EIF2AK2/PKR Antibody in Western Blot (WB)
          EIF2AK2/PKR Antibody in Western Blot (WB)
          EIF2AK2/PKR Antibody in Western Blot (WB)
          EIF2AK2/PKR Antibody in Western Blot (WB)
          EIF2AK2/PKR Antibody in Western Blot (WB)
          EIF2AK2/PKR Antibody in Western Blot (WB)
          EIF2AK2/PKR Antibody in Immunoprecipitation (IP)
          EIF2AK2/PKR Polyclonal Antibody

          Product Details

          18244-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 24 publications 24 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -

          RNA Immunoprecipitation (RIP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          EIF2AK2, PKR Fusion Protein Ag13010 (1-373 aa encoded by BC093676)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.21 mg/mL

          Amount

          31.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody is specific to EIF2AK2.

          Immunogen sequence: MAGDLSAGF FMEELNTYRQ KQGVVLKYQE LPNSGPPHDR RFTFQVIIDG REFPEGEGRS KKEAKNAAAK LAVEILNKEK KAVSPLLLTT TNSSEGLSMG NYIGLINRIA QKKRLTVNYE QCASGVHGPE GFHYKCKMGQ KEYSIGTGST KQEAKQLAAK LAYLQILSEE TSVKSDYLSS GSFATTCESQ SNSLVTSTLA SESSSEGDFS ADTSEINSNS DSLNSSSLLM NGLRNNQRKA KRSLAPRFDL PDMKETKYTV DKRFGMDFKE IELIGSGGFG QVFKAKHRID GKTYVIKRVK YNNEKAEREV KALAKLDHVN IVHYNGCWDG FDYDPETSDD SLESSDYDPE NSKNSSRSKT KCLFIQMEFC DKGT (1-373 aa encoded by BC093676)

          Target Information

          EIF2AK2 (PKR) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses. EIF2AK2 plays a key role in antiviral defense mediated through its direct activation by double-stranded RNA produced by viral infection. PKR plays a key role in IFN induced-innate antiviral response, virus-induced apoptosis, cell growth & differentiation. PKR induces apoptosis by up-regulating Fas expression and mediates FADD/Caspase-8 death signaling pathway. Upon viral infection, it functions as a dual protein, sequentially activating both cell survival & cell death pathways using kinase independent and dependent strategies. PRKR regulates multiple pathways which include NF-kappaB activation, p53, p38, & PDGF signaling pathway. PKR has been implicated in tumor suppression & malignancy. To evade the antiviral effects of PKR, viruses have evolved multiple mechanisms, such as the inhibition of PKR by the non-structural protein (NS1) of the influenza virus. More recently, PKR has been implicated in several neurodegenerative diseases including Alzheimer, Huntington, and amyotrophic lateral sclerosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: double stranded RNA activated protein kinase; dsRNA-activated kinase; EC 2.7.11.1; eIF-2 alpha; eIF-2A protein kinase 2; eIF2a Kinase; eukaryotic translation initiation factor 2 alpha kinase 2; Eukaryotic translation initiation factor 2-alpha kinase 2; IFN- type I-induced and dsRNA-activated kinase; IFN-induced and double-stranded RNA-activated kinase; Interferon-induced, double-stranded RNA-activated protein kinase; interferon-inducible elF2alpha kinase; Interferon-inducible RNA-dependent protein kinase; MGC126524; OTTHUMP00000201320; P1/eIF-2A protein kinase; p68 kinase; PKR; PRKR; protein kinase R; Protein kinase RNA-activated; protein kinase RNA-regulated; protein kinase, interferon inducible double stranded RNA dependent; Protein kinase, interferon-inducible double stranded RNA dependent; protein phosphatase 1, regulatory subunit 83; Serine/threonine-protein kinase TIK; T-cell viral integration site; Tyrosine-protein kinase EIF2AK2; unnamed protein product

          View more View less

          Gene Aliases: 2310047A08Rik; 4732414G15Rik; AI467567; AI747578; DYT33; EIF2AK1; EIF2AK2; LEUDEN; PKR; PPP1R83; PRKR; Tik

          View more View less

          UniProt ID: (Human) P19525, (Mouse) Q03963, (Rat) Q63184

          View more View less

          Entrez Gene ID: (Human) 5610, (Mouse) 19106, (Rat) 54287

          View more View less

          Function(s)
          nucleotide binding RNA binding double-stranded RNA binding protein kinase activity protein serine/threonine kinase activity eukaryotic translation initiation factor 2alpha kinase activity protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity protein binding ATP binding kinase activity transferase activity protein phosphatase regulator activity identical protein binding protein serine kinase activity transferase activity, transferring phosphorus-containing groups poly(A) RNA binding non-receptor serine/threonine protein kinase
          Process(es)
          positive regulation of cytokine production immune system process translation translational initiation regulation of translational initiation protein phosphorylation negative regulation of cell proliferation response to virus negative regulation of translation endoplasmic reticulum unfolded protein response positive regulation of chemokine production positive regulation of stress-activated MAPK cascade negative regulation of osteoblast proliferation cellular response to amino acid starvation response to interferon-alpha negative regulation of apoptotic process positive regulation of MAPK cascade negative regulation of viral genome replication innate immune response protein autophosphorylation positive regulation of NF-kappaB transcription factor activity defense response to virus antiviral innate immune response regulation of NLRP3 inflammasome complex assembly positive regulation of NIK/NF-kappaB signaling regulation of hematopoietic progenitor cell differentiation regulation of hematopoietic stem cell proliferation regulation of hematopoietic stem cell differentiation activation of MAPKK activity transcription, DNA-templated regulation of transcription, DNA-templated response to toxic substance phosphorylation response to lipopolysaccharide response to vitamin E positive regulation of apoptotic process response to mechanical stimulus response to organic substance peptidyl-tyrosine phosphorylation response to exogenous dsRNA
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6479c7f679-v472f:80/100.66.78.195:80.
          git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.50.0-Offline