Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • eIF4ENIF1 Antibodies

          Abnova

          EIF4ENIF1 Polyclonal Antibody, MaxPab™

          View all (11) eIF4ENIF1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite EIF4ENIF1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          EIF4ENIF1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          EIF4ENIF1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EIF4ENIF1 Antibody (H00056478-D01P) in WB

          Western Blot analysis of EIF4ENIF1 expression in transfected 293T cell line by EIF4ENIF1 MaxPab polyclonal antibody. Lane 1: EIF4ENIF1 transfected lysate(108.20 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EIF4ENIF1 Antibody in Western Blot (WB)

          Product Details

          H00056478-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          EIF4ENIF1 (AAH33028.1, 1 a.a. approximately 985 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MDRRSMGETE SGDAFLDLKK PPASKCPHRY TKEELLDIKE LPHSKQRPSC LSEKYDSDGV WDPEKWHASL YPASGRSSPV ESLKKELDTD RPSLVRRIVD PRERVKEDDL DVVLSPQRRS FGGGCHVTAA VSSRRSGSPL EKDSDGLRLL GGRRIGSGRI ISARTFEKDH RLSDKDLRDL RDRDRERDFK DKRFRREFGD SKRVFGERRR NDSYTEEEPE WFSAGPTSQS ETIELTGFDD KILEEDHKGR KRTRRRTASV KEGIVECNGG VAEEDEVEVI LAQEPAADQE VPRDAVLPEQ SPGDFDFNEF FNLDKVPCLA SMIEDVLGEG SVSASRFSRW FSNPSRSGSR SSSLGSTPHE ELERLAGLEQ AILSPGQNSG NYFAPIPLED HAENKVDILE MLQKAKVDLK PLLSSLSANK EKLKESSHSG VVLSVEEVEA GLKGLKVDQQ VKNSTPFMAE HLEETLSAVT NNRQLKKDGD MTAFNKLVST MKASGTLPSQ PKVSRNLESH LMSPAEIPGQ PVPKNILQEL LGQPVQRPAS SNLLSGLMGS LEPTTSLLGQ RAPSPPLSQV FQTRAASADY LRPRIPSPIG FTPGPQQLLG DPFQGMRKPM SPITAQMSQL ELQQAALEGL ALPHDLAVQA ANFYQPGFGK PQVDRTRDGF RNRQQRVTKS PAPVHRGNSS SPAPAASITS MLSPSFTPTS VIRKMYESKE KSKEEPASGK AALGDSKEDT QKASEENLLS SSSVPSADRD SSPTTNSKLS ALQRSSCSTP LSQANRYTKE QDYRPKATGR KTPTLASPVP TTPFLRPVHQ VPLVPHVPMV RPAHRLHPGL VQRMLAQGVH PQHLPSLLQT GVLPPGMDLS HLQGISGPIL GQPFYPLPAA SHPLLNPRPG TPLHLAMVQQ QLQRSVLHPP GSGSHAAAVS VQTTPQNVPS RSGLPHMHSQ LEHRPSQRSS SPVGLAKWFG SDVLQQPLPS MPAKVISVDE LEYRQ

          Target Information

          The protein encoded by this gene is a nucleocytoplasmic shuttle protein for the translation initiation factor eIF4E. This shuttle protein interacts with the importin alpha-beta complex to mediate nuclear import of eIF4E. It is predominantly cytoplasmic; its own nuclear import is regulated by a nuclear localization signal and nuclear export signals. Multiple transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 2610509L04Rik; 4E-T; 4E-transporter; bA247I13.2; eIF4E transporter; Eukaryotic translation initiation factor 4E nuclear import factor 1; Eukaryotic translation initiation factor 4E transporter; FLJ21601; FLJ26551; nucleocytoplasmic shuttling protein; OTTHUMP00000063276; unnamed protein product

          View more View less

          Gene Aliases: 4E-T; Clast4; EIF4ENIF1

          View more View less

          UniProt ID: (Human) Q9NRA8

          View more View less

          Entrez Gene ID: (Human) 56478

          View more View less

          Function(s)
          RNA binding mRNA binding nuclear export signal receptor activity protein binding kinase binding
          Process(es)
          translation regulation of translation protein import into nucleus protein transport negative regulation of translation stem cell population maintenance neuron differentiation gene silencing by RNA cytoplasmic mRNA processing body assembly miRNA mediated inhibition of translation negative regulation of neuron differentiation mRNA stabilization nuclear export positive regulation of nuclear-transcribed mRNA poly(A) tail shortening negative regulation of deadenylation-dependent decapping of nuclear-transcribed mRNA
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline