Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MNSQPQTRSP FAAGPRPPHH QFFQRPQIQP PRATIPNSSP SIRPGAQTPT AVYQANQHIM MVNHLPMPYP VPQGPQYCIP QYRHSGPPYV GPPQQYPVQP PGPGPFYPGP GPGDFPNAYG TPFYPSQPVY QSAPIMVPTQ QQPPPAKREK KTIRIRDPNQ GGKDITEEII SGGGSRNPTP PIGRPTSTPT PPQQLPSQVP EHSPVVYGTV ESAHLAASTP VTAASDQKQE EKPKPDPVLK SPSPVLRLVL SGEKKEQEGQ TSETTAIVSI AELPLPPSPT TVSSVARSTI AAPTSSALSS QPIFTTAIDD RCELSSPRED TIPIPSLTSC TETSDPLPTN ENDGDICKKP CSVAPNDIPL VSSTNLINEI NGVSEKLSAT ESIVEIVKQE VLPLTLELEI LENPPEEMKL ECIPAPITPS TVPSFPPTPP TPPASPPHTP VIVPAAATTV SSPSAAITVQ RVLEEDESIR TCLSEDAKEI QNKIEVEADG QTEEILDSQN LNSRRSPVPE TSNEC
The protein encoded by this gene is thought to be part of the eIF4F protein complex, which is involved in mRNA cap recognition and transport of mRNAs to the ribosome. Interestingly, a microRNA (miR-520c-3p) has been found that negatively regulates synthesis of the encoded protein, and this leads to a global decrease in protein translation and cell proliferation. Therefore, this protein is a key component of the anti-tumor activity of miR-520c-3p.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-4-gamma 3; eIF-4-gamma II; eIF4GII; Eukaryotic translation initiation factor 4 gamma 3; RP11-190H11.1
Gene Aliases: eIF-4G 3; eIF4G 3; EIF4G3; eIF4GII
UniProt ID: (Human) O43432
Entrez Gene ID: (Human) 8672
Molecular Function: translation factor translation initiation factor translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support