Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • eIF5B Antibodies

          Abnova

          EIF5B Polyclonal Antibody, MaxPab™

          View all (9) eIF5B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite EIF5B Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          EIF5B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          EIF5B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EIF5B Antibody (H00009669-B01P) in WB

          EIF5B MaxPab polyclonal antibody. Western Blot analysis of EIF5B expression in Jurkat. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EIF5B Antibody in Western Blot (WB)

          Product Details

          H00009669-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          EIF5B (AAH32639.1, 1 a.a. approximately 1220 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MGKKQKNKSE DSTKDDIDLD ALAAEIEGAG AAKEQEPQKS KGKKKKEKKK QDFDEDDILK ELEELSLEAQ GIKADRETVA VKPTENNEEE FTSKDKKKKG QKGKKQSFDD NDSEELEDKD SKSKKTAKPK VEMYSGSDDD DDFNKLPKKA KGKAQKSNKK WDGSEEDEDN SKKIKERSRI NSSGESGDES DEFLQSRKGQ KKNQKNKPGP NIESGNEDDD ASFKIKTVAQ KKAEKKERER KKRDEEKAKL RKLKEKEELE TGKKDQSKQK ESQRKFEEET VKSKVTVDTG VIPASEEKAE TPTAAEDDNE GDKKKKDKKK KKGEKEEKEK EKKKGPSKAT VKAMQEALAK LKEEEERQKR EEEERIKRLE ELEAKRKEEE RLEQEKRERK KQKEKERKER LKKEGKLLTK SQREARARAE ATLKLLQAQG VEVPSKDSLP KKRPIYEDKK RKKIPQQLES KEVSESMELC AAVEVMEQGV PEKEETPPPV EPEEEEDTED AGLDDWEAMA SDEETEKVEG NTVHIEVKEN PEEEEEEEEE EEEDEESEEE EEEEGESEGS EGDEEDEKVS DEKDSGKTLD KKPSKEMSSD SEYDSDDDRT KEERAYDKAK RRIEKRRLEH SKNVNTEKLR APIICVLGHV DTGKTKILDK LRHTHVQDGE AGGITQQIGA TNVPLEAINE QTKMIKNFDR ENVRIPGMLI IDTPGHESFS NLRNRGSSLC DIAILVVDIM HGLEPQTIES INLLKSKKCP FIVALNKIDR LYDWKKSPDS DVAATLKKQK KNTKDEFEER AKAIIVEFAQ QGLNAALFYE NKDPRTFVSL VPTSAHTGDG MGSLIYLLVE LTQTMLSKRL AHCEELRAQV MEVKALPGMG TTIDVILING RLKEGDTIIV PGVEGPIVTQ IRGLLLPPPM KELRVKNQYE KHKEVEAAQG VKILGKDLEK TLAGLPLLVA YKEDEIPVLK DELIHELKQT LNAIKLEEKG VYVQASTLGS LEALLEFLKT SEVPYAGINI GPVHKKDVMK ASVMLEHDPQ YAVNLAFDVR IERDAQEMAD SLGVRIFSAE IIYHLFDAFT KYRQDYKKQK QEEFKHIAVF PCKIKILPQY IFNSRDPIVM GVTVEAGQVK QGTPMCVPSK NFVDIGIVTS IEINHKQVDV AKKGQEVCVK IEPIPGESPK MFGRHFEATD ILVSKISRQS IDALKDWFRD EMQKSDWQLI VELKKVFEII

          Target Information

          Accurate initiation of translation in eukaryotes is complex and requires many factors, some of which are composed of multiple subunits. The process is simpler in prokaryotes which have only three initiation factors (IF1, IF2, IF3). Two of these factors are conserved in eukaryotes: the homolog of IF1 is eIF1A and the homolog of IF2 is eIF5B. This gene encodes eIF5B. Factors eIF1A and eIF5B interact on the ribosome along with other initiation factors and GTP to position the initiation methionine tRNA on the start codon of the mRNA so that translation initiates accurately.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DKFZp434I036; eIF 5B; eIF-5B; Eukaryotic translation initiation factor 5B; FLJ10524; Translation initiation factor IF-2; translation initiation factor IF2; unnamed protein product

          View more View less

          Gene Aliases: EIF5B; IF2; KIAA0741

          View more View less

          UniProt ID: (Human) O60841

          View more View less

          Entrez Gene ID: (Human) 9669

          View more View less

          Function(s)
          tRNA binding nucleotide binding RNA binding translation initiation factor activity GTPase activity protein binding GTP binding hydrolase activity metal ion binding
          Process(es)
          translation translational initiation regulation of translational initiation ribosome assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline