Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • EIN2 Antibodies

          Agrisera

          EIN2/Ethylene insensitive 2 Polyclonal Antibody

          View all (2) EIN2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite EIN2/Ethylene insensitive 2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          EIN2/Ethylene insensitive 2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          EIN2/Ethylene insensitive 2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EIN2/Ethylene insensitive 2 Antibody (AS12-1865) in WB

          40-60 µg of membrane protein (measured with Millipore Direct Detect spec. ) isolated following membrane extraction buffer protocol from Dong et al. 2008 (Plant Journal 53(2): 275-286) from 4 day old dark grown Arabidopsis thaliana seedlings, grown on MS plated with ACC or AVG in the plate, extracted with (10 m M Tris-HCl p H 7.5,150 m M Na Cl, 1 m M EDTA, 10% glycerol, 1% Triton X-100, and protease inhibito... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EIN2/Ethylene insensitive 2 Antibody in Western Blot (WB)

          Product Details

          AS12-1865

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000
          -
          Product Specifications

          Species Reactivity

          Plant

          Host/Isotype

          Rabbit

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH-conjugated peptide chosen from EIN2 of Arabidopsis thaliana.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Store lyophilized/reconstituted at -20°C; once reconstituted make aliquots to avoid repeated freeze-thaw cycles,Please remember to spin the tubes briefly prior to opening them to avoid any losses that might occur from material adhering to the cap or sides of the tube.

          For reconstitution add 50 µL of sterile water.

          Specific Species Reactivity: Arabidopsis thaliana

          Target Information

          Central factor in signaling pathways regulated by ethylene (ET) and involved in various processes including development, plant defense, senescence, nucleotide sugar flux, and tropisms (PubMed:27694846, PubMed:28874528). Necessary for ethylene-mediated gene regulation, and for the induction of some genes by ozone. Acts downstream of ET receptors, and upstream of ethylene regulated transcription factors. Required for cytokinin-mediated processes. Seems to be implicated in cross-talk between ET, jasmonate and other pathways. Probably not involved in iron uptake (PubMed:10381874, PubMed:12953109, PubMed:14973160, PubMed:15010611, PubMed:15272873, PubMed:7770519, PubMed:9351240). Has a short half-life and undergoes rapid proteasome-mediated turnover in the absence of ethylene (PubMed:19196655). Required for ethylene-induced EIN3 stabilization via proteasomal degradation of EBF1/EBF2 proteins (PubMed:20647342). Regulates the leaf senescence induced by methyl jasmonate, ethylene and abscisic acid (Ref.17, PubMed:15010611). Required during salt stress to confer resistance (PubMed:21631530).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ATEIN2; CKR1; CYTOKININ RESISTANT 1; Cytokinin-resistant protein AtCKR1; ENHANCED RESPONSE TO ABA3; ERA3; ETHYLENE INSENSITIVE 2; Ethylene-insensitive protein 2; F12E4.10; F12E4_10; ORE2; ORE3; ORESARA 2; ORESARA 3; PIR2; Protein ORESARA 3

          View more View less

          Gene Aliases: AT5G03280; ATEIN2; CKR1; CYTOKININ RESISTANT 1; EIN2; ENHANCED RESPONSE TO ABA3; ERA3; ETHYLENE INSENSITIVE 2; F12E4.10; F12E4_10; MOK16.19; ORE2; ORE3; ORESARA 2; ORESARA 3; PIR2

          View more View less

          UniProt ID: (Arabidopsis) Q9S814

          View more View less

          Entrez Gene ID: (Arabidopsis) 831889

          View more View less

          Function(s)
          mRNA binding manganese ion transmembrane transporter activity protein binding cadmium ion transmembrane transporter activity secondary carrier transporter
          Process(es)
          establishment of planar polarity response to molecule of bacterial origin response to osmotic stress response to oxidative stress cell death response to heat response to salt stress response to ethylene response to hormone response to jasmonic acid positive regulation of abscisic acid-activated signaling pathway jasmonic acid and ethylene-dependent systemic resistance, ethylene mediated signaling pathway ethylene-activated signaling pathway auxin polar transport phloem or xylem histogenesis regulation of stomatal movement leaf senescence sugar mediated signaling pathway negative regulation of defense response defense response to bacterium root hair cell differentiation defense response to fungus cell division defense response by callose deposition in cell wall
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-d4j86:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline