Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: PVSLRRGSHR LGSTSHFPQE KTHNHDPQPG DPNSRPSSPK PAQPALKMQV LYEFEARNPR ELTVVQGEKL EVLDHSKRWW LVKNEAGRSG YIPSNILEPL QPGTPGTQGQ SPSRVPMLRL SSRPEEVTDW LQAENFSTAT VRTLGSLTGS QLLRIRPGEL QMLCPQEAPR ILSRLEAVRR MLGISP*
This gene encodes a protein that is related to epidermal growth factor receptor pathway substrate 8 (EPS8), a substrate for the epidermal growth factor receptor. The function of this protein is unknown. Alternatively spliced transcript variants encoding different isoforms exist. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epidermal growth factor receptor kinase substrate 8-like protein 3; Epidermal growth factor receptor pathway substrate 8-related protein 3; EPS8 like 3; EPS8-like protein 3; EPS8-related protein 3; unnamed protein product
Gene Aliases: EPS8L3; EPS8R3; HYPT5; MUHH2
UniProt ID: (Human) Q8TE67
Entrez Gene ID: (Human) 79574
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support