Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MDPGKDKEG VPQPSGPPAR KKFVIPLDED EVPPGVAKPL FRSTQSLPTV DTSAQAAPQT YAEYAISQPL EGAGATCPTG SEPLAGETPN QALKPGAKSN SIIVSPRQRG NPVLKFVRNV PWEFGDVIPD YVLGQSTCAL FLSLRYHNLH PDYIHGRLQS LGKNFALRVL LVQVDVKDPQ QALKELAKMC ILADCTLILA WSPEEAGRYL ETYKAYEQKP ADLLMEKLEQ DFVSRVTECL TTVKSVNKTD SQTLLTTFGS LEQLIAASRE DLALCPGLGP QKVRALGKNP RSWGKERAPN KHNLRPQSFK VKKEPKTRHS GFRL (1-323 aa encoded by BC052813)
ERCC1 is a non-catalytic component of a structure-specific DNA repair endonuclease responsible for the 5'-incision during DNA repair. It is responsible, in conjunction with SLX4, for the first step in the repair of interstrand cross-links (ICL). It participates in the processing of anaphase bridge-generating DNA structures, which consist in incompletely processed DNA lesions arising during S or G2 phase, and can result in cytokinesis failure. ERCC1 is also required for homology-directed repair (HDR) of DNA double-strand breaks, in conjunction with SLX4. (Uniprot)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: clone 2B3; excision repair cross-complementation group 1; excision repair cross-complementing 1; excision repair cross-complementing rodent repair deficiency, complementation group 1; excision repair cross-complementing rodent repair deficiency, complementation group 1 (includes overlapping antisense sequence); unnamed protein product
Gene Aliases: COFS4; Ercc-1; RAD10; UV20
UniProt ID: (Human) P07992, (Mouse) P07903
Entrez Gene ID: (Human) 2067, (Rat) 292673, (Mouse) 13870
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support