Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • SMG5 Antibodies

          Abnova

          EST1B Polyclonal Antibody, MaxPab™

          View all (4) SMG5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite EST1B Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          EST1B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          EST1B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EST1B Antibody (H00023381-B01P) in WB

          Western Blot analysis of SMG5 expression in transfected 293T cell line by SMG5 MaxPab polyclonal antibody. Lane 1: EST1B transfected lysate(111.76 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EST1B Antibody in Western Blot (WB)

          Product Details

          H00023381-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          EST1B (NP_056142, 1 a.a. approximately 1016 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSQGPPTGES SEPEAKVLHT KRLYRAVVEA VHRLDLILCN KTAYQEVFKP ENISLRNKLR ELCVKLMFLH PVDYGRKAEE LLWRKVYYEV IQLIKTNKKH IHSRSTLECA YRTHLVAGIG FYQHLLLYIQ SHYQLELQCC IDWTHVTDPL IGCKKPVSAS GKEMDWAQMA CHRCLVYLGD LSRYQNELAG VDTELLAERF YYQALSVAPQ IGMPFNQLGT LAGSKYYNVE AMYCYLRCIQ SEVSFEGAYG NLKRLYDKAA KMYHQLKKCE TRKLSPGKKR CKDIKRLLVN FMYLQSLLQP KSSSVDSELT SLCQSVLEDF NLCLFYLPSS PNLSLASEDE EEYESGYAFL PDLLIFQMVI ICLMCVHSLE RAGSKQYSAA IAFTLALFSH LVNHVNIRLQ AELEEGENPV PAFQSDGTDE PESKEPVEKE EEPDPEPPPV TPQVGEGRKS RKFSRLSCLR RRRHPPKVGD DSDLSEGFES DSSHDSARAS EGSDSGSDKS LEGGGTAFDA ETDSEMNSQE SRSDLEDMEE EEGTRSPTLE PPRGRSEAPD SLNGPLGPSE ASIASNLQAM STQMFQTKRC FRLAPTFSNL LLQPTTNPHT SASHRPCVNG DVDKPSEPAS EEGSESEGSE SSGRSCRNER SIQEKLQVLM AEGLLPAVKV FLDWLRTNPD LIIVCAQSSQ SLWNRLSVLL NLLPAAGELQ ESGLALCPEV QDLLEGCELP DLPSSLLLPE DMALRNLPPL RAAHRRFNFD TDRPLLSTLE ESVVRICCIR SFGHFIARLQ GSILQFNPEV GIFVSIAQSE QESLLQQAQA QFRMAQEEAR RNRLMRDMAQ LRLQLEVSQL EGSLQQPKAQ SAMSPYLVPD TQALCHHLPV IRQLATSGRF IVIIPRTVID GLDLLKKEHP GARDGIRYLE AEFKKGNRYI RCQKEVGKSF ERHKLKRQDA DAWTLYKILD SCKQLTLAQG AGEEDPSGMV TIITGLPLDN PSVLSGPMQA ALQAAAHASV DIKDVLDFYK QWKEIG

          Target Information

          SMG5 plays a role in nonsense-mediated mRNA decay. SMG5 does not have RNase activity by itself. SMG5 promotes dephosphorylation of RENT1. SMG5 is necessary for TERT activity.SMG5 is involved in nonsense-mediated mRNA decay (Ohnishi et al., 2003 [PubMed 14636577]).[supplied by OMIM].

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: EST1 telomerase component homolog B; EST1-like protein B; EST1B; Est1p-like protein B; ever shorter telomeres 1B; FLJ34864; hSMG-5; LPTS interacting protein; LPTS-interacting protein; LPTS-RP1; Nonsense-mediated mRNA decay factor SMG5; protein SMG5; RP11-54H19.7; SMG-5 homolog; smg-5 homolog, nonsense mediated mRNA decay factor; unnamed protein product

          View more View less

          Gene Aliases: EST1B; KIAA1089; LPTS-RP1; LPTSRP1; SMG-5; SMG5

          View more View less

          UniProt ID: (Human) Q9UPR3

          View more View less

          Entrez Gene ID: (Human) 23381

          View more View less

          Function(s)
          protein binding ubiquitin protein ligase binding telomeric DNA binding histone deacetylase binding protein phosphatase 2A binding telomerase RNA binding
          Process(es)
          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay mRNA export from nucleus regulation of telomere maintenance regulation of telomere maintenance via telomerase regulation of dephosphorylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline