Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Antibody detects endogenous levels of total EXOSC10.
About 50% of patients with polymyositis/scleroderma (PM-Scl) overlap syndrome are reported to have autoantibodies to a nuclear/nucleolar particle termed PM-Scl. Exosome component 10 (EXOSC10), also named autoantigen PM/Scl-2, is the 100 kDa antigen component of PM-Scl and is recognized by most sera of PM-Scl patients. EXOSC10 is strongly enriched in the nucleolus and a small amount has been found in cytoplasm supporting the existence of a nucleolar RNA exosome complex form. As a putative catalytic component ofthe RNA exosome complex which has 3'->5' exoribonuclease activity, EXOSC10 participates in a multitude of cellular RNA processing and degradation events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: autoantigen PM-SCL; autoantigen PM/Scl 2 homolog; P100 polymyositis-scleroderma overlap syndrome associated autoantigen homolog; P100 polymyositis-scleroderma overlap syndrome-associated autoantigen; PM/Scl; polymyositis/scleroderma autoantigen 100 kDa; polymyositis/scleroderma autoantigen 2; polymyositis/scleroderma autoantigen 2 homolog; unnamed protein product
Gene Aliases: p2; p3; p4; PM-Scl; PM/Scl-100; PM/Scl100; PMSCL; PMSCL2; RRP6; Rrp6p
UniProt ID: (Human) Q01780, (Rat) D4A1X2, (Mouse) P56960
Entrez Gene ID: (Human) 5394, (Rat) 313707, (Mouse) 50912
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support