Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • EXT1 Antibodies

          Abnova

          EXT1 Polyclonal Antibody, MaxPab™

          View all (6) EXT1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite EXT1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          EXT1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          EXT1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EXT1 Antibody (H00002131-D01P) in ICC/IF

          Immunofluorescence of purified MaxPab antibody to EXT1 on HeLa cell. Antibody concentration is 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EXT1 Antibody in Immunocytochemistry (ICC/IF)
          EXT1 Antibody in Western Blot (WB)
          EXT1 Antibody in Western Blot (WB)
          EXT1 Polyclonal Antibody, MaxPab™

          Product Details

          H00002131-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          EXT1 (NP_000118.2, 1 a.a. approximately 746 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MQAKKRYFIL LSAGSCLALL FYFGGLQFRA SRSHSRREEH SGRNGLHHPS PDHFWPRFPD ALRPFVPWDQ LENEDSSVHI SPRQKRDANS SIYKGKKCRM ESCFDFTLCK KNGFKVYVYP QQKGEKIAES YQNILAAIEG SRFYTSDPSQ ACLFVLSLDT LDRDQLSPQY VHNLRSKVQS LHLWNNGRNH LIFNLYSGTW PDYTEDVGFD IGQAMLAKAS ISTENFRPNF DVSIPLFSKD HPRTGGERGF LKFNTIPPLR KYMLVFKGKR YLTGIGSDTR NALYHVHNGE DVVLLTTCKH GKDWQKHKDS RCDRDNTEYE KYDYREMLHN ATFCLVPRGR RLGSFRFLEA LQAACVPVML SNGWELPFSE VINWNQAAVI GDERLLLQIP STIRSIHQDK ILALRQQTQF LWEAYFSSVE KIVLTTLEII QDRIFKHISR NSLIWNKHPG GLFVLPQYSS YLGDFPYYYA NLGLKPPSKF TAVIHAVTPL VSQSQPVLKL LVAAAKSQYC AQIIVLWNCD KPLPAKHRWP ATAVPVVVIE GESKVMSSRF LPYDNIITDA VLSLDEDTVL STTEVDFAFT VWQSFPERIV GYPARSHFWD NSKERWGYTS KWTNDYSMVL TGAAIYHKYY HYLYSHYLPA SLKNMVDQLA NCEDILMNFL VSAVTKLPPI KVTQKKQYKE TMMGQTSRAS RWADPDHFAQ RQSCMNTFAS WFGYMPLIHS QMRLDPVLFK DQVSILRKKY RDIERL

          Target Information

          Hereditary multiple exostoses (HME) is an autosomal dominant disorder characterized by the formation of exostoses (EXT), which are cartilage-capped bony protuberances mainly located on long bones. Two proteins associated with EXT, EXT1 and EXT2, form homo/heteromeric complexes in vivo, which leads to the accumulation of both proteins in the Golgi apparatus. EXT1 and EXT2 are endoplasmic reticulum-localized type II transmembrane glycoproteins that possess, or are tightly associated with, glycosyltransferase activities involved in the polymerization of the glycosaminoglycan, heparan sulfate (HS). EXT2 is a protein that harbors the D-glucuronyl (GlcA) and N-acetyl-D-glucosaminyl (GlcNAc) transferase activities required for biosynthesis of HS. EXT1 rescues defective HS biosynthesis and elevates low GlcA and GlcNAc transferase levels in mutated cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: exostoses (multiple) 1; Exostosin glycosyltransferase 1; Exostosin-1; Glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N- acetylglucosaminyltransferase; glucuronosyl-N-acetylglucosaminyl-proteoglycan/N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Heparan sulfate co-polymerase subunit EXT1; Langer-Giedion syndrome chromosome region; Multiple exostoses protein 1; N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase; putative tumor suppressor protein EXT1; putative tumour suppressor/hereditary multiple exostoses candidate gene; unnamed protein product

          View more View less

          Gene Aliases: EXT; EXT1; LGCR; LGS; TRPS2; TTV

          View more View less

          UniProt ID: (Human) Q16394

          View more View less

          Entrez Gene ID: (Human) 2131

          View more View less

          Function(s)
          protein binding acetylglucosaminyltransferase activity glucuronosyltransferase activity transferase activity transferase activity, transferring glycosyl groups heparan sulfate N-acetylglucosaminyltransferase activity protein homodimerization activity metal ion binding protein heterodimerization activity glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase activity
          Process(es)
          polysaccharide biosynthetic process cell morphogenesis ossification endochondral ossification blood vessel remodeling chondrocyte differentiation glandular epithelial cell differentiation hematopoietic progenitor cell differentiation hypersensitivity heart field specification chondrocyte hypertrophy endochondral bone growth glycosaminoglycan biosynthetic process protein glycosylation vacuole organization cell adhesion cell surface receptor signaling pathway smoothened signaling pathway gastrulation axon guidance brain development endoderm development mesoderm development regulation of blood pressure cell proliferation fibroblast growth factor receptor signaling pathway response to virus response to light intensity gene expression regulation of tumor necrosis factor-mediated signaling pathway neural crest cell differentiation heparan sulfate proteoglycan biosynthetic process stem cell division antigen processing and presentation optic nerve development olfactory bulb development hair cycle process protein catabolic process proteoglycan biosynthetic process collagen fibril organization heparin metabolic process heparin biosynthetic process BMP signaling pathway hair follicle morphogenesis neuron projection development glomerular basement membrane development cell adhesion mediated by integrin social behavior synaptic transmission, glutamatergic multicellular organism growth chondrocyte proliferation limb joint morphogenesis dendritic cell migration lymphocyte adhesion to endothelial cell of high endothelial venule fluid transport wound healing vasodilation fear response ossification involved in bone maturation cellular response to fibroblast growth factor stimulus cell fate commitment bone resorption embryonic morphogenesis sebaceous gland development chondroitin sulfate proteoglycan metabolic process multicellular organismal water homeostasis leukocyte tethering or rolling sulfation sodium ion homeostasis heart contraction canonical Wnt signaling pathway hematopoietic stem cell differentiation bone morphogenesis endochondral bone morphogenesis cartilage development involved in endochondral bone morphogenesis epithelium development epithelial tube branching involved in lung morphogenesis mesenchyme development smoothened signaling pathway involved in lung development developmental growth involved in morphogenesis sweat gland development hematopoietic stem cell homeostasis motor behavior perichondral bone morphogenesis stomach development protein-containing complex assembly dendrite self-avoidance response to growth factor response to heparin vocalization behavior basement membrane organization cellular response to BMP stimulus glomerular visceral epithelial cell differentiation embryonic skeletal joint development lymphocyte migration into lymphoid organs hematopoietic stem cell migration to bone marrow cellular response to virus bone growth tight junction organization mesenchymal cell differentiation involved in bone development cranial skeletal system development response to leukemia inhibitory factor
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline