PA1-30239 detects Enterococcus from bacteria samples.
PA1-30239 has been successfully used in immunofluorescence applications.
The PA1-30239 immunogen is: Enterococcus faecium.
Enterococci form part of the lactic acid bacteria family and are the third most important bacterial genus in hospital infections, especially Enterococcus (E.) faecium. In food they can spoil processed meats but they are equally important for ripening and aroma development of certain traditional cheeses and sausages and are used as human probiotics. However, they possesses a broad spectrum antibiotic resistance which has important implications to human health and disease.