Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • Ephrin B2 Antibodies

          Invitrogen

          Ephrin B2 Polyclonal Antibody

          View all (20) Ephrin B2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Ephrin B2 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (38)
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 38

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Ephrin B2 Antibody (PA5-106933) in IHC (P)

          Immunohistochemistry analysis of paraffin-embedded Ephrin B2 in human meningeal carcinomatosis(MC) tissue sections. Antigen retrieval was performed using citrate buffer. Samples were blocked with blocking buffer (1.5 hr, 22°C), incubated with Ephrin B2 polyclonal antibody (Product # PA5-106933) using a dilution of 1:200 (1.5 hr, 22°C), followed by HRP conjugated goat anti-rabbit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ephrin B2 Antibody in Western Blot (WB)
          Ephrin B2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Ephrin B2 Polyclonal Antibody

          Product Details

          PA5-106933

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human EFNB2(Accession P52799), corresponding to amino acid residues Y304-V333.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2854597

          Product Specific Information

          Antibody detects endogenous levels of total Ephrin B2.

          Target Information

          This gene encodes a member of the ephrin (EPH) family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. This gene encodes an EFNB class ephrin which binds to the EPHB4 and EPHA3 receptors.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Efnb2; EPH-related receptor tyrosine kinase ligand 5; HTK ligand; ligand of eph-related kinase 5; ligand of the eph-related receptor tyrosine kinases hek and elk; membrane protein; MGC126226; MGC126227; MGC126228; unnamed protein product

          View more View less

          Gene Aliases: ELF-2; ephrin-B2; Epl5; EPLG5; Htk-L; HTKL; LERK-5; LERK5; NLERK-1

          View more View less

          UniProt ID: (Human) P52799, (Mouse) P52800

          View more View less

          Entrez Gene ID: (Human) 1948, (Rat) 306636, (Mouse) 13642

          View more View less

          Function(s)
          receptor binding protein binding ephrin receptor binding receptor agonist activity transmembrane receptor protein tyrosine kinase activator activity
          Process(es)
          angiogenesis lymph vessel development cell migration involved in sprouting angiogenesis cell adhesion axon guidance positive regulation of cell proliferation organ morphogenesis negative regulation of keratinocyte proliferation negative regulation of neuron projection development T cell costimulation adherens junction organization ephrin receptor signaling pathway blood vessel morphogenesis anatomical structure formation involved in morphogenesis venous blood vessel morphogenesis regulation of chemotaxis cellular response to lipopolysaccharide nephric duct morphogenesis presynapse assembly regulation of postsynaptic membrane neurotransmitter receptor levels regulation of postsynaptic neurotransmitter receptor internalization positive regulation of aorta morphogenesis positive regulation of leukocyte adhesion to arterial endothelial cell positive regulation of cardiac muscle cell differentiation anatomical structure morphogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-dg9d8:80/100.66.76.64:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0