Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ERK1/2 Antibodies

          Rockland

          Erk1/2 Polyclonal Antibody

          View all (105) ERK1/2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Erk1/2 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          Erk1/2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Erk1/2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Erk1/2 Antibody (100-401-F25) in ICC/IF

          Immunofluorescence Microscopy of Rabbit anti-ERK1/2 antibody. Tissue: backskin sections of transgenic mice. Fixation: 0.5% PFA. Antigen retrieval: not required. Primary antibody: ERK1/2 antibody at 10 µg/mL for 1 hr at RT. Secondary antibody: Fluorescein rabbit secondary antibody at 1:10,000 for 45 min at RT. Localization: ERK1/2 is cytoplasmic. Staining: ERK1/2 as green fluorescent signal. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Erk1/2 Antibody in Immunocytochemistry (ICC/IF)
          Erk1/2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          100-401-F25

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Bovine, Chicken, Fruit fly, Human, Mouse, Sheep, Rat, Xenopus

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Erk 1/2 Antibody was produced from whole rabbit serum prepared by repeated immunizations with a synthetic peptide, corresponding to Erk1 MAP kinase with the CGG spacer group added and the synthetic peptide coupled to KLH.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS with 50% glycerol

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          ERK1 and ERK2 are widely expressed and are involved in the regulation of meiosis, mitosis, and postmitotic functions in differentiated cells. Many different stimuli, including growth factors, cytokines, virus infection, ligands for heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors and transforming agents, activate the ERK1 and ERK2 pathways. When growth factors bind to the receptor tyrosine kinase, Ras interacts with Raf, the serine/threonine protein kinase and activates it as well. Once actived, Raf phosphorylates serine residue in 2 further kinases, MEK1/2, which in turn phosphorylates tyrosine/threonine in extracellular-signal regulated kinase (ERK) 1/2. Upon activation, the ERKs either phosphorylate a number of cytoplasmic targets or migrate to the nucleus, where they phosphorylate and activate a number of transcription factors such as c-Fos and Elk-1.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CG12559-PA; CG12559-PC; CG12559-PD; CG12559-PE; CG12559-PG; CG12559-PH; CG12559-PI; Diphospho-ERK; dp-ERK; dpERK; drosophila ERK; enhancer of seven in absentia 7; ERK; Erk MAP kinase; ERK-2; ERK/rolled; erk1 erk2; Erk1; cancer susceptibility protein; ERK1; MAP kinase; mitogen-activated protein kinase; ERK1b; ERK1c; Erk2; cancer susceptibility protein; ERT1; extracellular signal-regulated kinase; extracellular signal-regulated kinase 1; extracellular signal-regulated kinase 2; extracellular signal-regulated kinase A; extracellular signal-related kinase 1; Extracellular-regulated kinase A; extracellular-signal-regulated kinase 2; Extracellular-signal-related kinase A; insulin-stimulated MAP2 kinase; MAP kinase; MAP kinase 1; MAP kinase 2; MAP kinase 3; MAP-kinase; MAPK; MAPK 1; MAPK 2; MAPK 3; microtubule-associated protein 2 kinase; mitogen activated protein kinase; mitogen-activated 3; mitogen-activated protein kinase; mitogen-activated protein kinase 1; mitogen-activated protein kinase 1a; mitogen-activated protein kinase 2; p42; p42 MAP Kinase; p42-MAPK; p44 MAP kinase; p44/42 MAPK; phospho-ERK; pp42/MAP kinase; protein serine/threonine kinase ERK1b; protein tyrosine kinase ERK2; RE08694p; rl-PA; rl-PC; rl-PD; rl-PE; rl-PG; rl-PH; rl-PI; roll; sevenmaker; unnamed protein product

          View more View less

          Gene Aliases: 12559; 9030612K14Rik; BcDNA:RE08694; CG12559; CG18732; CT34260; CT39192; D-ERK; DERK; DERK-A; dm-dpERK; Dmel\CG12559; Dmel_CG12559; DmERK; DmERK-A; DmERKA; DmMAPK; dp-ERK; DpERK; dpERK1; dpMAPK; Dsor2; E(sina)7; EC2-1; EK2-1; ERK; ERK-1; ERK-2; ERK-A; Erk/Map kinase; ERK1; ERK1/2; ERK2; ERKA; ERT1; ERT2; Esrk1; EY2-2; GroupII; HS44KDAP; HUMKER1A; l(2)41Ac; l(2R)EMS45-39; MAP-k; MAPK; MAPK1; mapk1-a; mapk1-b; mapk1a; MAPK2; MNK1; mpk1; Mtap2k; NS13; p-ERK; p38; p40; p41; p41mapk; p42-MAPK; P42MAPK; p44; p44-ERK1; p44-MAPK; P44ERK1; P44MAPK; pERK; pMAPK; PRKM1; PRKM2; PRKM3; RL; rl/ERK; rl/MAPK; rll; SEM; SR2-1; Su(Raf)2B; XELAEV_18010534mg; xp42

          View more View less

          UniProt ID: (Bovine) P46196, (Human) P28482, (Human) P27361, (Rat) P63086, (Mouse) P63085, (Mouse) Q63844, (Rat) P21708, (Xenopus) P26696, (Fruit fly) P40417

          View more View less

          Entrez Gene ID: (Bovine) 327672, (Human) 5594, (Sheep) 101115485, (Bovine) 531391, (Chicken) 373953, (Sheep) 101123008, (Human) 5595, (Rat) 116590, (Mouse) 26413, (Mouse) 26417, (Rat) 50689, (Xenopus) 398985, (Fruit fly) 3354888

          View more View less

          Function(s)
          phosphotyrosine binding protein serine/threonine kinase activity MAP kinase kinase activity ATP binding RNA polymerase II carboxy-terminal domain kinase activity phosphatase binding identical protein binding protein kinase activity MAP kinase activity protein binding kinase activity protein serine kinase activity DNA-binding transcription factor binding double-stranded DNA binding protein kinase binding mitogen-activated protein kinase kinase kinase binding scaffold protein binding transcription factor binding non-receptor serine/threonine protein kinase
          Process(es)
          activation of MAPK activity protein phosphorylation apoptotic process cellular response to DNA damage stimulus cell cycle positive regulation of gene expression positive regulation of peptidyl-threonine phosphorylation peptidyl-serine phosphorylation peptidyl-threonine phosphorylation cytosine metabolic process regulation of ossification regulation of cellular pH thyroid gland development regulation of protein stability lipopolysaccharide-mediated signaling pathway positive regulation of telomere maintenance via telomerase mammary gland epithelial cell proliferation cellular response to amino acid starvation cellular response to reactive oxygen species response to nicotine ERBB signaling pathway outer ear morphogenesis response to exogenous dsRNA negative regulation of cell differentiation thymus development T cell receptor signaling pathway B cell receptor signaling pathway stress-activated MAPK cascade positive regulation of telomerase activity Bergmann glial cell differentiation long-term synaptic potentiation face development lung morphogenesis trachea formation labyrinthine layer blood vessel development cardiac neural crest cell development involved in heart development ERK1 and ERK2 cascade response to epidermal growth factor cellular response to cadmium ion cellular response to tumor necrosis factor cellular response to granulocyte macrophage colony-stimulating factor stimulus cellular response to dopamine positive regulation of telomere capping MAPK cascade regulation of transcription from RNA polymerase II promoter chemotaxis signal transduction cell surface receptor signaling pathway epidermal growth factor receptor signaling pathway chemical synaptic transmission learning or memory insulin receptor signaling pathway positive regulation of macrophage chemotaxis Schwann cell development positive regulation of telomere maintenance regulation of stress-activated MAPK cascade intracellular signal transduction ERBB2-ERBB3 signaling pathway myelination positive regulation of skeletal muscle tissue regeneration positive regulation of cholesterol biosynthetic process positive regulation of smoothened signaling pathway insulin-like growth factor receptor signaling pathway regulation of cytoskeleton organization interleukin-34-mediated signaling pathway chemokine-mediated signaling pathway caveolin-mediated endocytosis regulation of Golgi inheritance positive regulation of macrophage proliferation positive regulation of neuroinflammatory response regulation of early endosome to late endosome transport negative regulation of T cell mediated immune response to tumor cell phosphorylation sensory perception of pain BMP signaling pathway positive regulation of cyclase activity peptidyl-tyrosine autophosphorylation signal transduction in response to DNA damage positive regulation of transcription from RNA polymerase II promoter modulation of synaptic transmission positive regulation of ERK1 and ERK2 cascade interleukin-1-mediated signaling pathway cellular response to mechanical stimulus negative regulation of cholesterol efflux negative regulation of TORC1 signaling heart development positive regulation of cell proliferation response to toxic substance organ morphogenesis neural crest cell development diadenosine tetraphosphate biosynthetic process positive regulation of cell migration response to estradiol response to lipopolysaccharide response to testosterone cellular response to platelet-derived growth factor stimulus response to cocaine positive regulation of protein import into nucleus response to hydrogen peroxide response to estrogen estrous cycle positive regulation of translation positive regulation of transcription, DNA-templated decidualization regulation of intracellular pH positive regulation of cardiac muscle cell proliferation cellular response to methionine cellular response to cAMP cellular response to epidermal growth factor stimulus cellular response to peptide hormone stimulus cellular response to prostaglandin E stimulus cellular response to lipid cellular response to toxic substance response to alcohol cellular response to insulin-like growth factor stimulus placenta development autophagosome assembly negative regulation of autophagy positive regulation of autophagy cellular response to nutrient levels negative regulation of proteasomal ubiquitin-dependent protein catabolic process TORC1 signaling protein stabilization negative regulation of T cell activation cartilage development protein-containing complex assembly positive regulation of xenophagy arachidonic acid metabolic process mitotic cell cycle terminal region determination terminal branching, open tracheal system R7 cell fate commitment imaginal disc-derived wing vein specification imaginal disc-derived wing morphogenesis leg disc proximal/distal pattern formation metamorphosis torso signaling pathway determination of adult lifespan fibroblast growth factor receptor signaling pathway cellular response to starvation regulation of gene expression negative regulation of macroautophagy lymph gland plasmatocyte differentiation lymph gland crystal cell differentiation tracheal pit formation in open tracheal system sevenless signaling pathway positive regulation of cell size positive regulation of photoreceptor cell differentiation dorsal appendage formation vascular endothelial growth factor receptor signaling pathway behavioral response to ethanol positive regulation of sequence-specific DNA binding transcription factor activity defense response to virus branching involved in open tracheal system development cellular response to arsenic-containing substance positive regulation of wound healing regulation of heart morphogenesis regulation of response to drug
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch