Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MSFSGKYQL QSQENFEAFM KAIGLPEELI QKGKDIKGVS EIVQNGKHFK FTITAGSKVI QNEFTVGEEC ELETMTGEKV KTVVQLEGDN KLVTTFKNIK SVTELNGDII TNTMTLGDIV FKRISKRI (1-127 aa encoded by BC032801)
FABP1 encodes the fatty acid binding protein found in liver. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP1 and FABP6 (the ileal fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 14 kDa selenium-binding protein; FABP1; fatty acid binding protein; fatty acid binding protein (FABP); fatty acid binding protein 1, liver; fatty acid-binding protein 1; L-FABP; liver fatty acid binding protein p14; liver-type fatty acid-binding protein; squalene- and sterol-carrier protein; sterol carrier protein; unnamed protein product; Z-protein
Gene Aliases: FABPL; Fabplg; L-FABP; p14; SCP; SCP.
UniProt ID: (Human) P07148, (Rat) P02692, (Mouse) P12710
Entrez Gene ID: (Human) 2168, (Rat) 24360, (Mouse) 14080
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support