Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAFDSTWKV DRSENYDKFM EKMGVNIVKR KLAAHDNLKL TITQEGNKFT VKESSAFRNI EVVFELGVTF NYNLADGTEL RGTWSLEGNK LIGKFKRTDN GNELNTVREI IGDELVQTYV YEGVEAKRIF KKD (1-132 aa encoded by BC069617)
The FABP2 gene encodes the intestinal fatty acid-binding protein 2 (I-FABP), which plays a significant role in lipid metabolism by facilitating the transport and intracellular trafficking of long-chain fatty acids in enterocytes, the absorptive cells in the intestine. FABP2 is involved in the uptake, metabolism, and transfer of fatty acids across cellular membranes, contributing to digestion and absorption processes in the intestine. This protein is crucial for efficient lipid metabolism, influencing fatty acid oxidation, and energy balance. Variations in the FABP2 gene have been associated with metabolic traits, such as insulin sensitivity, lipid profiles, and obesity, suggesting its impact on metabolic health. FABP2's function in intestinal lipid absorption and metabolism makes it a subject of interest in studies related to dietary effects on health and the development of metabolic disorders. Understanding FABP2's role is essential for insights into nutritional influences on health and possible therapeutic targets for metabolic diseases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FABP2; FABPI; fatty acid binding protein (FABP); fatty acid binding protein 1; fatty acid binding protein 2, intestinal; I-FABP; intestinal FABP; intestinal fatty acid binding protein; intestinal fatty acid binding protein 2; intestinal-type fatty acid-binding protein; MGC133132; putative; unnamed protein product
Gene Aliases: FABP; FABPI; I-FABP
UniProt ID: (Human) P12104, (Rat) P02693, (Mouse) P55050
Entrez Gene ID: (Human) 2169, (Rat) 25598, (Mouse) 14079
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support