Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FAM160B1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 160 member B1; family with sequence similarity 160, member B1; FHF complex subunit HOOK interacting protein 2A; FHIP2A; protein FAM160B1
Gene Aliases: AI450540; bA106M7.3; FAM160B1; FHIP2A; KIAA1600; mKIAA1600; RGD1306116
UniProt ID: (Human) Q5W0V3, (Mouse) Q8CDM8
Entrez Gene ID: (Human) 57700, (Mouse) 226252, (Rat) 361774
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support