Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: SSRGREQAVR KSEKERMREY QRELEEREEK LKKRPLLFER VAQKNARMAA EKHYSNTLKA LGISDEFVSK KGQSGKVLEY FNNQETKSVT EDKESFNEEE KIEERENGEE NYFIDTNSQD SYKEKDEANE ESEEEKSVEE SH
This gene belongs to the FAM161 family. It is expressed mainly in the retina. Mouse studies suggested that this gene is involved in development of retinal progenitors during embryogenesis, and that its activity is restricted to mature photoreceptors after birth. Mutations in this gene cause autosomal recessive retinitis pigmentosa-28. Alternatively spliced transcript variants have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FAM161A, centrosomal protein; family with sequence similarity 161 member A; Protein FAM161A; retinitis pigmentosa 28 (autosomal recessive); unnamed protein product
Gene Aliases: FAM161A; RGD1304999; RP28
UniProt ID: (Human) Q3B820, (Rat) Q6AY14
Entrez Gene ID: (Human) 84140, (Rat) 289833
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support