Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MSSTFWAFM ILASLLIAYC SQLAAGTCEI VTLDRDSSQP RRTIARQTAR CACRKGQIAG TTRARPACVD ARIIKTKQWC DMLPCLEGEG CDLLINRSGW TCTQPGGRIK TTTVS (1-114 aa encoded by BC039396)
Acts as a chemokine-like protein by regulating cell proliferation and migration through activation of G protein-coupled receptors (GPCRs), such as S1PR2 and FPR2. Stimulates chemotactic migration of macrophages mediated by the MAPK3/ERK1 and AKT1 pathway. Blocks TNFSF11/RANKL-induced osteoclast formation from macrophages by inhibiting up-regulation of osteoclast fusogenic and differentiation genes. Stimulation of macrophage migration and inhibition of osteoclast formation is mediated via GPCR FPR2. Acts as an adipokine by negatively regulating vascular smooth muscle cell (VSMC) proliferation and migration in response to platelet-derived growth factor stimulation via GPCR S1PR2 and G protein GNA12/GNA13-transmitted RHOA signaling . Inhibits injury-induced cell proliferation and neointima formation in the femoral arteries
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 19 (chemokine (C-C motif)-like), member A5; family with sequence similarity 19 member A5, C-C motif chemokine like; family with sequence similarity 19, member A5; protein FAM19A5; TAFA protein 5; unnamed protein product
Gene Aliases: FAM19A5; QLLK5208; RGD1562115; TAFA-5; Tara-5; UNQ5208
UniProt ID: (Human) Q7Z5A7, (Mouse) Q91WE9, (Rat) M0R7X9
Entrez Gene ID: (Human) 25817, (Mouse) 106014, (Rat) 500915
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support