Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FAM62A Antibodies

          Abnova

          FAM62A Polyclonal Antibody, MaxPab™

          View all (12) FAM62A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite FAM62A Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          FAM62A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FAM62A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FAM62A Antibody (H00023344-B01P) in WB

          FAM62A MaxPab polyclonal antibody. Western Blot analysis of FAM62A expression in HepG2. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FAM62A Antibody in Western Blot (WB)
          FAM62A Polyclonal Antibody, MaxPab™

          Product Details

          H00023344-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          FAM62A (NP_056107.1, 1 a.a. approximately 1104 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MERSPGEGPS PSPMDQPSAP SDPTDQPPAA HAKPDPGSGG QPAGPGAAGE ALAVLTSFGR RLLVLIPVYL AGAVGLSVGF VLFGLALYLG WRRVRDEKER SLRAARQLLD DEEQLTAKTL YMSHRELPAW VSFPDVEKAE WLNKIVAQVW PFLGQYMEKL LAETVAPAVR GSNPHLQTFT FTRVELGEKP LRIIGVKVHP GQRKEQILLD LNISYVGDVQ IDVEVKKYFC KAGVKGMQLH GVLRVILEPL IGDLPFVGAV SMFFIRRPTL DINWTGMTNL LDIPGLSSLS DTMIMDSIAA FLVLPNRLLV PLVPDLQDVA QLRSPLPRGI IRIHLLAARG LSSKDKYVKG LIEGKSDPYA LVRLGTQTFC SRVIDEELNP QWGETYEVMV HEVPGQEIEV EVFDKDPDKD DFLGRMKLDV GKVLQASVLD DWFPLQGGQG QVHLRLEWLS LLSDAEKLEQ VLQWNWGVSS RPDPPSAAIL VVYLDRAQDL PLKKGNKEPN PMVQLSIQDV TQESKAVYST NCPVWEEAFR FFLQDPQSQE LDVQVKDDSR ALTLGALTLP LARLLTAPEL ILDQWFQLSS SGPNSRLYMK LVMRILYLDS SEICFPTVPG CPGAWDVDSE NPQRGSSVDA PPRPCHTTPD SQFGTEHVLR IHVLEAQDLI AKDRFLGGLV KGKSDPYVKL KLAGRSFRSH VVREDLNPRW NEVFEVIVTS VPGQELEVEV FDKDLDKDDF LGRCKVRLTT VLNSGFLDEW LTLEDVPSGR LHLRLERLTP RPTAAELEEV LQVNSLIQTQ KSAELAAALL SIYMERAEDL PLRKGTKHLS PYATLTVGDS SHKTKTISQT SAPVWDESAS FLIRKPHTES LELQVRGEGT GVLGSLSLPL SELLVADQLC LDRWFTLSSG QGQVLLRAQL GILVSQHSGV EAHSHSYSHS SSSLSEEPEL SGGPPHITSS APELRQRLTH VDSPLEAPAG PLGQVKLTLW YYSEERKLVS IVHGCRSLRQ NGRDPPDPYV SLLLLPDKNR GTKRRTSQKK RTLSPEFNER FEWELPLDEA QRRKLDVSVK SNSSFMSRER ELLGKVQLDL AETDLSQGVA RWYDLMDNKD KGSS

          Target Information

          May play a role as calcium-regulated intrinsic membrane protein. Target protein is an enriched in adipocyte plasma membranes and high density microsome fractions.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: E-Syt1; extended synaptotagmin like protein 1; extended synaptotagmin protein 1; Extended synaptotagmin-1; family with sequence similarity 62 (C2 domain containing), member A; Membrane-bound C2 domain-containing protein; multiple C2 domain protein; unnamed protein product

          View more View less

          Gene Aliases: ESYT1; FAM62A; KIAA0747; MBC2

          View more View less

          UniProt ID: (Human) Q9BSJ8

          View more View less

          Entrez Gene ID: (Human) 23344

          View more View less

          Function(s)
          calcium ion binding protein binding calcium-dependent phospholipid binding lipid binding phosphatidylethanolamine binding phosphatidylcholine binding phosphatidylinositol binding identical protein binding metal ion binding intermembrane phospholipid transfer activity
          Process(es)
          lipid transport phospholipid transport endoplasmic reticulum-plasma membrane tethering intermembrane lipid transfer
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0