Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • FCRL3 Antibodies

          Abnova

          FCRL3 Polyclonal Antibody, MaxPab™

          View all (10) FCRL3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite FCRL3 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          FCRL3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FCRL3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FCRL3 Antibody (H00115352-B01P) in WB

          Western Blot analysis of FCRL3 expression in transfected 293T cell line by FCRL3 MaxPab polyclonal antibody. Lane 1: FCRL3 transfected lysate(81.62 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FCRL3 Antibody in Western Blot (WB)
          FCRL3 Polyclonal Antibody, MaxPab™

          Product Details

          H00115352-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          FCRL3 (ABM86598.1, 1 a.a. approximately 742 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MLLWLLLLIL TPGREQSGVA PKAVLLLNPP WSTAFKGEKV ALICSSISHS LAQGDTYWYH DEKLLKIKHD KIQITEPGNY QCKTRGSSLS DAVHVEFSPD WLILQALHPV FEGDNVILRC QGKDNKNTHQ KVYYKDGKQL PNSYNLEKIT VNSVSRDNSK YHCTAYRKFY ILDIEVTSKP LNIQVQELFL HPVLRASSST PIEGSPMTLT CETQLSPQRP DVQLQFSLFR DSQTLGLGWS RSPRLQIPAM WTEDSGSYWC EVETVTHSIK KRSLRSQIRV QRVPVSNVNL EIRPTGGQLI EGENMVLICS VAQGSGTVTF SWHKEGRVRS LGRKTQRSLL AELHVLTVKE SDAGRYYCAA DNVHSPILST WIRVTVRIPV SHPVLTFRAP RAHTVVGDLL ELHCESLRGS PPILYRFYHE DVTLGNSSAP SGGGASFNLS LTAEHSGNYS CDADNGLGAQ HSHGVSLRVT VPVSRPVLTL RAPGAQAVVG DLLELHCESL RGSFPILYWF YHEDDTLGNI SAHSGGGASF NLSLTTEHSG NYSCEADNGL GAQHSKVVTL NVTGTSRNRT GLTAAGITGL VLSILVLAAA AALLHYARAR RKPGGLSATG TSSHSPSECQ EPSSSRPSRI DPQEPTHSKP LAPMELEPMY SNVNPGDSNP IYSQIWSIQH TKENSANCPM MHQEHEELTV LYSELKKTHP DDSAGEASSR GRAHEEDDEE NYENILNPRK NKVQDFPCLC NT

          Target Information

          This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein contains immunoreceptor-tyrosine activation motifs and immunoreceptor-tyrosine inhibitory motifs in its cytoplasmic domain and may play a role in regulation of the immune system. Mutations in this gene have been associated with rheumatoid arthritis, autoimmune thyroid disease, and systemic lupus erythematosus. Alternative splicing results in multiple transcript variants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD307c; Fc receptor homolog 3; Fc receptor-like protein 3; FcR-like protein 3; FcRH3; hIFGP3; IFGP family protein 3; Ig-like superfamily receptor; Ig-superfamily protein SPAP2a; Ig-superfamily protein SPAP2b; Ig-superfamily protein SPAP2c; Ig-superfamily protein SPAP2d; Ig-superfamily protein SPAP2e; Immune receptor translocation-associated protein 3; immunoglobulin superfamily receptor translocation associated protein 3; IRTA3; MAIA; SH2 domain-containing phosphatase anchor protein 2; unnamed protein product

          View more View less

          Gene Aliases: CD307c; FCRH3; FCRL3; IFGP3; IRTA3; MAIA; SPAP2

          View more View less

          UniProt ID: (Human) Q96P31

          View more View less

          Entrez Gene ID: (Human) 115352

          View more View less

          Function(s)
          transmembrane signaling receptor activity protein binding kinase binding phosphatase binding
          Process(es)
          negative regulation of immunoglobulin production immune response cell surface receptor signaling pathway single fertilization positive regulation of B cell proliferation regulation of toll-like receptor 9 signaling pathway positive regulation of MAPK cascade regulation of B cell differentiation negative regulation of B cell receptor signaling pathway regulation of B cell activation regulation of calcium ion import
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline