Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • FEM1C Antibodies

          Proteintech

          FEM1C Polyclonal Antibody

          View all (5) FEM1C antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite FEM1C Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (15)
          FEM1C Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FEM1C Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 15

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FEM1C Antibody (14037-1-AP) in ICC/IF

          Immunofluorescent analysis of HeLa cells using 14037-1-AP (FEM1C antibody) at dilution of 1:25 and Rhodamine-Goat anti-Rabbit IGG. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FEM1C Antibody in Immunocytochemistry (ICC/IF)
          FEM1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FEM1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FEM1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FEM1C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FEM1C Antibody in Western Blot (WB)
          FEM1C Antibody in Western Blot (WB)
          FEM1C Antibody in Western Blot (WB)
          FEM1C Antibody in Western Blot (WB)
          FEM1C Antibody in Western Blot (WB)
          FEM1C Antibody in Western Blot (WB)
          FEM1C Antibody in Western Blot (WB)
          FEM1C Antibody in Western Blot (WB)
          FEM1C Antibody in Western Blot (WB)
          FEM1C Antibody in Western Blot (WB)
          FEM1C Polyclonal Antibody

          Product Details

          14037-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          FEM1C Fusion Protein Ag5099 (315-617 aa encoded by BC028369)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.13 mg/mL

          Amount

          19.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody specifically recognizes the 69 kDa FEM1C protein.

          Immunogen sequence: PDEMRMQAL LIRERILGPS HPDTSYYIRY RGAVYADSGN FKRCINLWKY ALDMQQSNLD PLSPMTASSL LSFAELFSFM LQDRAKGLLG TTVTFDDLMG ILCKSVLEIE RAIKQTQCPA DPLQLNKALS IILHLICLLE KVPCTLEQDH FKKQTIYRFL KLHPRGKNNF SPLHLAVDKN TTCVGRYPVC KFPSLQVTAI LIECGADVNV RDSDDNSPLH IAALNNHPDI MNLLIKSGAH FDATNLHKQT ASDLLDEKEI AKNLIQPINH TTLQCLAARV IVNHRIYYKG HIPEKLETFV SLHR (315-617 aa encoded by B C028369)

          Target Information

          Probable component of an E3 ubiquitin-protein ligase complex, in which it may act as a substrate recognition subunit.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FEM1-gamma; FEM1c; Protein fem-1 homolog C; similar to Caenorhabditis elegans sex-determination protein FEM-1; unnamed protein product

          View more View less

          Gene Aliases: 2610312A07Rik; 3632443A22Rik; AI646567; EUROIMAGE686608; EUROIMAGE783647; FEM1A; FEM1C; KIAA1785

          View more View less

          UniProt ID: (Human) Q96JP0, (Mouse) Q8CEF1

          View more View less

          Entrez Gene ID: (Human) 56929, (Mouse) 240263

          View more View less

          Function(s)
          protein binding protein binding, bridging involved in substrate recognition for ubiquitination molecular_function
          Process(es)
          protein ubiquitination proteasome-mediated ubiquitin-dependent protein catabolic process ubiquitin-dependent protein catabolic process via the C-end degron rule pathway biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline