Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FGF10 Antibodies

          Invitrogen

          FGF10 Polyclonal Antibody, Biotin, PeproTech®

          View all (13) FGF10 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite FGF10 Polyclonal Antibody, Biotin, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          FGF10 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          FGF10 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FGF10 Antibody (500-P151GBT-25UG) in ELISA

          Sandwich ELISA: To detect Human FGF-10 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of FGF10 Polyclonal Antibody, Biotin (Product # 500-P151GBT-500UG) is required. This biotinylated polyclonal antibody, in conjunction with PeproTech FGF10 Polyclonal Antibody (Product # 500-P151G-1MG) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human FGF-10. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FGF10 Antibody in ELISA (ELISA)
          FGF10 Polyclonal Antibody, Biotin, PeproTech®

          Product Details

          500-P151GBT-25UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.2 µg/mL
          -

          ELISA (ELISA)

          0.25-1.0 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Goat

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived Recombinant Human FGF-10
          3D Epitope / Immunogen

          Conjugate

          Biotin Biotin Biotin

          Form

          Lyophilized

          Concentration

          0.1-1.0 mg/mL

          Amount

          25 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_2929494

          Product Specific Information

          AA Sequence of recombinant protein: MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEI GVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS.

          Preparation: Produced from sera of goats immunized with highly pure Recombinant Human FGF-10. Anti-Human FGF-10-specific antibody was purified by affinity chromatography and then biotinylated.

          Sandwich ELISA: To detect Human FGF-10 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human FGF-10 (500-P151G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human FGF-10.

          Western Blot: To detect hFGF-10 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hFGF-10 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

          500-P151GBT-1MG will be provided as 2 x 500 µg

          Target Information

          FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This protein is also implicated to be a primary factor in the process of wound healing.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FGF; FGF-10; FGF10; Fibroblast growth factor; Fibroblast growth factor 10; fibroblast growth factor 10; FGF10; Keratinocyte growth factor 2; keratinocyte growth factor-2; KGF2; produced by fibroblasts of urinary bladder lamina propria

          View more View less

          Gene Aliases: FGF10; LADD3

          View more View less

          UniProt ID: (Human) O15520

          View more View less

          Entrez Gene ID: (Human) 2255

          View more View less

          Function(s)
          fibroblast growth factor receptor binding type 2 fibroblast growth factor receptor binding protein binding growth factor activity heparin binding chemoattractant activity
          Process(es)
          establishment of mitotic spindle orientation mitotic cell cycle angiogenesis metanephros development organ induction mesonephros development endothelial cell proliferation positive regulation of endothelial cell proliferation blood vessel remodeling metanephros morphogenesis chemotaxis cell-cell signaling determination of left/right symmetry salivary gland development salivary gland morphogenesis cell proliferation positive regulation of cell proliferation fibroblast growth factor receptor signaling pathway epidermis development regulation of smoothened signaling pathway embryonic pattern specification organ morphogenesis regulation of gene expression positive regulation of epithelial cell migration positive regulation of keratinocyte proliferation Wnt signaling pathway pituitary gland development neurogenesis actin cytoskeleton organization cell differentiation positive regulation of Wnt signaling pathway lung development regulation of cell migration embryonic genitalia morphogenesis epithelial cell differentiation negative regulation of epithelial cell differentiation thyroid gland development otic vesicle formation positive regulation of vascular endothelial growth factor receptor signaling pathway pancreas development hair follicle morphogenesis embryonic camera-type eye development response to estradiol response to lipopolysaccharide positive regulation of ATPase activity lacrimal gland development regulation of activin receptor signaling pathway protein localization to cell surface somatic stem cell population maintenance limb morphogenesis organ growth wound healing tissue regeneration inner ear morphogenesis odontogenesis of dentin-containing tooth muscle cell fate commitment positive regulation of MAPK cascade keratinocyte proliferation positive regulation of DNA repair positive regulation of Notch signaling pathway positive regulation of transcription, DNA-templated positive regulation of mitotic cell cycle positive regulation of transcription from RNA polymerase II promoter positive regulation of Ras protein signal transduction regulation of saliva secretion vascular endothelial growth factor receptor signaling pathway fibroblast proliferation positive regulation of fibroblast proliferation lung alveolus development blood vessel morphogenesis spleen development thymus development embryonic digestive tract morphogenesis digestive tract development embryonic digestive tract development organ formation epidermis morphogenesis semicircular canal morphogenesis branching morphogenesis of an epithelial tube female genitalia morphogenesis male genitalia morphogenesis positive regulation of lymphocyte proliferation epithelial cell proliferation urothelial cell proliferation positive regulation of urothelial cell proliferation positive regulation of epithelial cell proliferation positive regulation of peptidyl-tyrosine phosphorylation white fat cell differentiation positive chemotaxis induction of positive chemotaxis smooth muscle cell differentiation positive regulation of keratinocyte migration radial glial cell differentiation positive regulation of epithelial cell proliferation involved in wound healing limb development limb bud formation lung morphogenesis lung epithelium development lung saccule development bronchiole morphogenesis epithelial tube branching involved in lung morphogenesis branching involved in salivary gland morphogenesis bud outgrowth involved in lung branching bud elongation involved in lung branching mesenchymal-epithelial cell signaling involved in lung development Type II pneumocyte differentiation prostatic bud formation respiratory system development mammary gland specification fibroblast growth factor receptor signaling pathway involved in mammary gland specification mammary gland bud formation submandibular salivary gland formation epithelial cell proliferation involved in salivary gland morphogenesis regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling branch elongation involved in salivary gland morphogenesis semicircular canal fusion mesenchymal cell differentiation involved in lung development secretion by lung epithelial cell involved in lung growth lung proximal/distal axis specification tear secretion white fat cell proliferation positive regulation of white fat cell proliferation ERK1 and ERK2 cascade positive regulation of ERK1 and ERK2 cascade Harderian gland development positive regulation of hair follicle cell proliferation stem cell proliferation positive regulation of canonical Wnt signaling pathway extrinsic apoptotic signaling pathway in absence of ligand positive regulation of G1/S transition of mitotic cell cycle fibroblast growth factor receptor apoptotic signaling pathway negative regulation of stem cell proliferation positive regulation of stem cell proliferation negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline