Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAKVFSFIL VTTALTMGRE ISALEDCAQE QMRLRAQVRL LETRVKQQQV KIKQLLQENE VQFLDKGDEN TVIDLGSKRQ YADCSEIFND GYKLSGFYKI KPLQSPAEFS VYCDMSDGGG WTVIQRRSDG SENFNRGWKD YENGFGNFVQ KHGEYWLGNK NLHFLTTQED YTLKIDLADF EKNSRYAQYK NFKVGDEKNF YELNIGEYSG TAGDSLAGNF HPEVQWWASH QRMKFSTWDR DHDNYEGNCA EEDQSGWWFN RCHSANLNGV YYSGPYTAKT DNGIVWYTWH GWWYSLKSVV MKIRPNDFIP NVI (1-312 aa encoded by BC007047)
Fibrinogen-like 1 is a member of the fibrinogen family. This protein is homologous to the carboxy terminus of the fibrinogen beta- and gamma- subunits which contains the four conserved cysteines of fibrinogens and fibrinogen related proteins. However, this protein lacks the platelet-binding site, cross-linking region and a thrombin-sensitive site which are necessary for fibrin clot formation. This protein may play a role in the development of hepatocellular carcinomas. Four alternatively spliced transcript variants encoding the same protein exist for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fibrinogen-related protein 1; fibronigen-like protein 1; hepassocin; hepatocellular carcinoma-related sequence; hepatocyte-derived fibrinogen-related protein 1; liver fibrinogen-related protein 1; liver fibrinogen-related protein-1; MFIRE-1; mfrep-1; MGC12455; OTTHUMP00000122468; unnamed protein product
Gene Aliases: Frep1; HFREP1; HP-041; HPS; LFIRE-1; LFIRE1; Mfire1
UniProt ID: (Human) Q08830, (Rat) Q5M8C6, (Mouse) Q71KU9
Entrez Gene ID: (Human) 2267, (Rat) 246186, (Mouse) 234199
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support