FIGURE: 1 / 2
Recommended positive controls: HepG2, mouse liver.
Predicted reactivity: Mouse (86%), Rat (86%), Bovine (92%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Fibrinogen-like 1 is a member of the fibrinogen family. This protein is homologous to the carboxy terminus of the fibrinogen beta- and gamma- subunits which contains the four conserved cysteines of fibrinogens and fibrinogen related proteins. However, this protein lacks the platelet-binding site, cross-linking region and a thrombin-sensitive site which are necessary for fibrin clot formation. This protein may play a role in the development of hepatocellular carcinomas. Four alternatively spliced transcript variants encoding the same protein exist for this gene.