    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • LAMP2 (CD107b) Antibodies
          Proteintech
          CD107b/LAMP2 Recombinant Rabbit Monoclonal Antibody (H4B4), FITC Plus
          View all (70) LAMP2 (CD107b) antibodies
          Tech Support
          Cite CD107b/LAMP2 Recombinant Rabbit Monoclonal Antibody (H4B4), FITC Plus
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          CD107b/LAMP2 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD107b/LAMP2 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          CD107b/LAMP2 Antibody (FITC-65520) in Flow
          1x10^6 human PBMCs were intracellularly stained with 5 µL FITC Plus Anti-Human CD107b Rabbit Recombinant Antibody (FITC-65520, Clone: H4B4) or 5 µL FITC Plus Anti-Human CD107b Mouse Monoclonal Antibody (FITC-65053, Clone: H4B4). Cells were fixed and permeabilized with Transcription Factor Staining Buffer Kit (PF00011). Cells were treated with FC Receptor Block prior to staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD107b/LAMP2 Antibody in Flow Cytometry (Flow)
          CD107b/LAMP2 Antibody in Flow Cytometry (Flow)
          CD107b/LAMP2 Antibody in Flow Cytometry (Flow)
          CD107b/LAMP2 Recombinant Rabbit Monoclonal Antibody (H4B4), FITC Plus
          Product Details
          FITC-65520
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          proprietary
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          H4B4
          Immunogen
          Human adherent peripheral blood cells
          View immunogen
          Conjugate
          FITC Plus FITC Plus FITC Plus
          • Unconjugated
          • CoraLite Plus 647
          • Request custom conjugation
          Excitation/Emission Max
          495/524 nm
          Form
          Liquid
          Concentration
          5 µL/Test
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 0.5% BSA
          Contains
          0.09% sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Wet ice
          Target Information
          LAMP2 (Lysosome-associated membrane glycoprotein 2, CD107b) is a member of a family of membrane glycoproteins. LAMP2 provides selectins with carbohydrate ligands, and may play a role in tumor cell metastasis, and have a function in the protection, maintenance, and adhesion of the lysosome. Prior to posttranslational modification, LAMP2 is approximately 46 kDa polypeptide. Mature, functional LAMP2 is extensively glycosylated with a variety of different N-linked and O-linked oligosaccharides with a total molecular weight of about 100-130 kDa. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail. LAMP2 is used to monitor a very selective form of autophagy called chaperone-mediated autophagy. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail. The LAMP protein has 3 variant forms: LAMP-2A, -2B, and 2C. LAMP2, together with CD107a/LAMP-1, is a major constituent of lysosomal membrane. The LAMP proteins are involved in lysosome biogenesis and are required for fusion of lysosomes with phagosomes, and LAMP2 is an important regulator in successful phagosomal maturation. LAMP2 deficiency causes an accumulation of autophagosomes in many tissues leading to cardiomyopathy and myopathy (Danon's disease).
          Bioinformatics
          Protein Aliases: CD107 antigen-like family member B; CD107b; Igp96; LAMP-2; LAMP-2 Isoform 2A; LAMP-2 Variant A; LGP-96; lysosomal-associated membrane protein 2; Lysosome-associated membrane glycoprotein 2; RP23-193O17.2
          View more View less
          Gene Aliases: CD107b; LAMP-2; LAMP2; LAMPB; LGP110
          View more View less
          UniProt ID: (Human) P13473
          View more View less
          Entrez Gene ID: (Human) 3920
          View more View less
          Function(s)
          protein binding enzyme binding protein domain specific binding membrane trafficking regulatory protein
          Process(es)
          platelet degranulation protein import regulation of protein stability muscle cell cellular homeostasis protein stabilization chaperone-mediated autophagy
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0