Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody can recognize all the 3 isoforms of FLCN (38-45 kDa,64 kDa,20-26 kDa).
Immunogen sequence: MNAIVALCH FCELHGPRTL FCTEVLHAPL PQGDGNEDSP GQGEQAEEEE GGIQMNSRMR AHSPAEGASV ESSSPGPKKS DMCEGCRSLA AGHPGYISHD KETSIKYVSH QHPSHPQLFS IVRQACVRSL SCEVCPGREG PIFFGDEQHG FVFSHTFFIK DSLARGFQRW YSIITIMMDR IYLINSWPFL LGKVRGIIDE LQGKALKVFE AEQFGCPQRA QRMNTAFTPF LHQRNGNAAR SLTSLTSDDN LWACLHTSFA WLLKACGSRL TEKLLEGAPT EDTLVQMEKL AGEAGVLLPG PWPGWPWGGT SCLLSWQESL REGNAALNQP RTSLREAHPP ISV (1-342 aa encoded by BC015687)
This gene encodes a mitochondrial protein that is characterized by a poly-proline rich region. A chicken homolog of this protein promotes mitochondrial fission and the mouse homolog protects cells from oxidative stress. A related pseudogene of this gene is found on chromosome X.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BHD skin lesion fibrofolliculoma protein; birt-Hogg-Dube syndrome protein; birt-Hogg-Dube syndrome protein homolog; unnamed protein product
Gene Aliases: B430214A04Rik; BHD; DENND8B; FLCL
UniProt ID: (Human) Q8NFG4, (Mouse) Q8QZS3, (Rat) Q76JQ2
Entrez Gene ID: (Human) 201163, (Mouse) 216805, (Rat) 303185
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support