Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • FLIP Antibodies

          Rockland

          FLIP Polyclonal Antibody

          View all (18) FLIP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite FLIP Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          FLIP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FLIP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FLIP Antibody (600-401-BF3) in ICC/IF

          Immunocytochemistry of Rabbit anti-FLIP antibody. Cell Type: HeLa cells. Fixation: formalin fixed paraffin embedded. Antigen retrieval: not required. Primary antibody: anti-FLIP antibody at 5 µg/mL for 1 hr at RT. Secondary antibody: Peroxidase rabbit secondary antibody at 1:10,000 for 45 min at RT. Localization: FLIP is cytoplasmic. Staining: FLIP as precipitated brown signal with hematoxylin purple nuclear counterstain. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FLIP Antibody in Immunocytochemistry (ICC/IF)
          FLIP Antibody in Immunocytochemistry (ICC/IF)
          FLIP Antibody in Western Blot (WB)
          FLIP Polyclonal Antibody

          Product Details

          600-401-BF3

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-2 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          5 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          20 µg/mL
          -

          Flow Cytometry (Flow)

          Assay-dependent
          -

          ELISA (ELISA)

          1:10,000-1:20,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Anti-FLIP antibody was prepared from whole rabbit serum produced by repeated immunizations with a peptide corresponding to amino acids near the N-terminus of human FLIP.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          FLIP (CFLAR) is an apoptosis regulator protein which functions as a crucial link between cell survival and cell death pathways in mammalian cells. It acts as an inhibitor of TNFRSF6 mediated apoptosis. A proteolytic fragment (p43) is likely retained in the death-inducing signaling complex (DISC) thereby blocking further recruitment and processing of caspase-8 at the complex. Full length and shorter isoforms have been shown either to induce apoptosis or to reduce TNFRSF-triggered apoptosis. Lacks enzymatic (caspase) activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: c-FLIP; CASH; CASP8 and FADD-like apoptosis regulator; Caspase homolog; Caspase-eight-related protein; Caspase-like apoptosis regulatory protein; caspase-related inducer of apoptosis; Casper; Cellular FLICE-like inhibitory protein; CLARP; DED-caspase homologue; FADD-like antiapoptotic molecule 1; FLAME-1; FLIP-L; apoptosis inhibitor; FLIP-S; apoptosis inhibitor; I-FLICE; Inhibitor of FLICE; MACH-related inducer of toxicity; MRIT; testis secretory sperm-binding protein Li 233m; unnamed protein product; Usurpin; usurpin beta

          View more View less

          Gene Aliases: 2310024N18Rik; A430105C05Rik; c-FLIP; c-FLIPL; c-FLIPR; c-FLIPS; CASH; CASP8AP1; Casper; CFLAR; cFLIP; CLARP; ENSMUSG00000072980; FLAME; FLAME-1; FLAME1; FLIP; Gm9845; I-FLICE; MRIT

          View more View less

          UniProt ID: (Human) O15519, (Mouse) O35732

          View more View less

          Entrez Gene ID: (Human) 8837, (Rat) 117279, (Mouse) 12633

          View more View less

          Function(s)
          protease binding cysteine-type endopeptidase activity death receptor binding protein binding enzyme activator activity cysteine-type peptidase activity macromolecular complex binding peptidase activator activity protein complex binding death effector domain binding protein heterodimerization activity cysteine-type endopeptidase activity involved in apoptotic signaling pathway protease
          Process(es)
          response to hypoxia proteolysis apoptotic process skeletal muscle tissue development extrinsic apoptotic signaling pathway via death domain receptors negative regulation of cardiac muscle cell apoptotic process positive regulation of neuron projection development skeletal muscle atrophy regulation of skeletal muscle satellite cell proliferation skeletal myofibril assembly macrophage differentiation cellular response to insulin stimulus response to testosterone regulation of apoptotic process negative regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling skeletal muscle tissue regeneration positive regulation of neuron apoptotic process regulation of necroptotic process positive regulation of ERK1 and ERK2 cascade cellular response to epidermal growth factor stimulus cellular response to estradiol stimulus cellular response to hypoxia cellular response to dexamethasone stimulus cellular response to nitric oxide positive regulation of glomerular mesangial cell proliferation negative regulation of myoblast fusion negative regulation of extrinsic apoptotic signaling pathway via death domain receptors positive regulation of extracellular matrix organization negative regulation of reactive oxygen species biosynthetic process negative regulation of cellular response to transforming growth factor beta stimulus negative regulation of hepatocyte apoptotic process negative regulation of epithelial cell apoptotic process positive regulation of hepatocyte proliferation negative regulation of extrinsic apoptotic signaling pathway positive regulation of peptidase activity positive regulation of catalytic activity negative regulation of cysteine-type endopeptidase activity involved in apoptotic process negative regulation of necroptotic process activation of cysteine-type endopeptidase activity involved in apoptotic process apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-d4j86:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline