Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • FNDC3A Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.

          Abnova

          FNDC3A Polyclonal Antibody, MaxPab™

          View all (7) FNDC3A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Tech Support
          Datasheet
          Certificates
          Protocols
          Tech Support
          Datasheet
          Certificates
          Protocols
          Tech Support

          Cite FNDC3A Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          FNDC3A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FNDC3A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FNDC3A Antibody (H00022862-B01P) in WB

          Western Blot analysis of FNDC3A expression in transfected 293T cell line (H00022862-T01) by FNDC3A MaxPab polyclonal antibody.

          Lane1:FNDC3A transfected lysate(124.74 KDa).
          Lane2:Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FNDC3A Antibody in Western Blot (WB)
          FNDC3A Antibody in Western Blot (WB)
          FNDC3A Polyclonal Antibody, MaxPab™

          Product Details

          H00022862-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          FNDC3A (ENSP00000347952, 1 a.a. approximately 1134 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          50 µg

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MMSPNGSVPP IYVPPGYAPQ VIEDNGVRRV VVVPQAPEFH PGSHTVLHRS PHPPLPGFIP VPTMMPPPPR HMYSPVTGAG DMTTQYMPQY QSSQVYGDVD AHSTHGRSNF RDERSSKTYE RLQKKLKDRQ GTQKDKMSSP PSSPQKCPSP INEHNGLIKG QIAGGINTGS AKIKSGKGKG GTQVDTEIEE KDEETKAFEA LLSNIVKPVA SDIQARTVVL TWSPPSSLIN GETDESSVPE LYGYEVLISS TGKDGKYKSV YVGEETNITL NDLKPAMDYH AKVQAEYNSI KGTPSEAEIF TTLSCEPDIP NPPRIANRTK NSLTLQWKAP SDNGSKIQNF VLEWDEGKGN GEFCQCYMGS QKQFKITKLS PAMGCKFRLS ARNDYGTSGF SEEVLYYTSG CAPSMPASPV LTKAGITWLS LQWSKPSGTP SDEGISYILE MEEETSGYGF KPKYDGEDLA YTVKNLRRST KYKFKVIAYN SEGKSNPSEV VEFTTCPDKP GIPVKPSVKG KIHSHSFKIT WDPPKDNGGA TINKYVVEMA EGSNGNKWEM IYSGATREHL CDRLNPGCFY RLRVYCISDG GQSAVSESLL VQTPAVPPGP CLPPRLQGRP KAKEIQLRWG PPLVDGGSPI SCYSVEMSPI EKDEPREVYQ GSEVECTVSS LLPGKTYSFR LRAANKMGFG PFSEKCDITT APGPPDQCKP PQVTCRSATC AQVNWEVPLS NGTDVTEYRL EWGGVEGSMQ ICYCGPGLSY EIKGLSPATT YYCRVQALSV VGAGPFSEVV ACVTPPSVPG IVTCLQEISD DEIENPHYSP STCLAISWEK PCDHGSEILA YSIDFGDKQS LTVGKVTSYI INNLQPDTTY RIRIQALNSL GAGPFSHMIK LKTKPLPPDP PRLECVAFSH QNLKLKWGEG TPKTLSTDSI QYHLQMEDKN GRFVSLYRGP CHTYKVQRLN ESTSYKFCIQ ACNEAGEGPL SQEYIFTTPK SVPAALKAPK IEKVNDHICE ITWECLQPMK GDPVIYSLQV MLGKDSEFKQ IYKGPDSSFR YSSLQLNCEY RFRVCAIRQC QDSLGHQDLV GPYSTTVLFI SQRTEPPAST NRDTVESTRT RRALSDEQCA AVILVLFAFF SILIAFIIQY FVIK

          Target Information

          FNDC3A gene ontology annotations related to this gene include cell-cell adhesion; fertilization; Sertoli cell development; spermatid development.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline