Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEEQREALRK IIKTLAVKNE EIQSFIYSLK QMLLNVEANS AKVQEDLEAE FQSLFSLLEE LKEGMLMKIK QDRASRTYEL QNQLAACTRA LESSEELLET ANQTLQAMDS EDFPQAAKQI KDGVTMAPAF RLSLKAKVSD NMSHLMVDFA QERQMLQALK FLPVPSAPVI DLAESLVADN CVTLVWRMPD EDSKIDHYVL EYRRTNFEGP PRLKEDQPWM VIEGIRQTEY TLTGLKFDMK YMNFRVKACN KAVAGEFSEP VTLETPAFMF RLDASTSHQN LRVDDLSVEW DAMGGKVQDI KAREKDGKGR TASPINSPAR GTPSPKRMPS GRGGRDRFTA ESYTVLGDTL IDGGEHYWEV RYEPDSKAFG VGVAYRSLGR FEQLGKTAAS WCLHVNNWLQ VSFTAKHANK VKVLDAPVPD CLGVHCDFHQ GLLSFYNART KQVLHTFKTR FTQPLLPAFT VWCGSFQVTT GLQVPSAVRC LQKRGSATSS SNTSLT
FSD1 encodes a centrosome associated protein that is characterized by an N-terminal coiled-coil region downstream of B-box (BBC) domain, a central fibronectin type III domain, and a C-terminal repeats in splA and RyR (SPRY) domain. The encoded protein associates with a subset of microtubules and may be involved in the stability and organization of microtubules during cytokinesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fibronectin type 3 and SPRY (spla, ryanodine) domain containing (with coiled-coil motif) 1; fibronectin type 3 and SPRY domain containing 1; FSD1; sp1a/ryanodine domain; contains a coiled-coil motif; microtubule-associated protein GLFND; MID1-related protein 1; midline 1-related protein 1; unnamed protein product
Gene Aliases: GLFND; MIR1
UniProt ID: (Human) Q9BTV5
Entrez Gene ID: (Human) 79187
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support