PA1-27575 detects FTSJ3 from human samples.
PA1-27575 may be successfully used in Western blot applications.
The immunogen is a synthetic peptide corresponding to human Putative rRNA methyltransferase 3.
Although the function of this gene is not known, the existence of this gene is supported by mRNA and EST data. A possible function of the encoded protein can be inferred from amino acid sequence similarity to the E.coli FtsJ protein and to a mouse protein possibly involved in embryogenesis.