Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Positive Control - WB: Rat Kidney Tissue, SW620 whole cell, A549 whole cell. IHC: mouse intestine tissue, rat intestine tissue, human mammary cancer tissue.
FUT1 is a Golgi stack membrane protein that is involved in the creation of a precursor of the H antigen, which is required for the final step in the soluble A and B antigen synthesis pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a-1, 2 fucosyltransferase (H antigen); alpha 1,2-fucosyltransferase A; alpha 12-fucosyltransferase; alpha(1,2) fucosyltransferase 1; alpha(1,2)FT 1; alpha12-fucosyltransferase a; beta-galactoside alpha-1,2-fucosyltransferase; blood group H alpha 2-fucosyltransferase; fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase); galactoside 2-alpha-L-fucosyltransferase; galactoside 2-alpha-L-fucosyltransferase 1; galactoside 2-L-fucosyltransferase enzyme; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1; H antigen; H glycosyltransferase; H glycosytransferase; H transferase; nonfunctional fucosyltransferase 1; type 1 galactoside alpha-(1,2)-fucosyltransferase FUT1; type 2 galactoside alpha-(1,2)-fucosyltransferase FUT1; unnamed protein product
Gene Aliases: FTA; Futa; H; HH; HSC; MFUT-1
UniProt ID: (Human) P19526, (Rat) Q10980, (Mouse) O09160
Entrez Gene ID: (Human) 2523, (Rat) 81919, (Mouse) 14343
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support