Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
This gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. The function of this protein is unknown, although it may play a role in mammalian hair follicle development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: frizzled 3; frizzled 3, seven transmembrane spanning receptor; frizzled class receptor 3; frizzled family receptor 3; frizzled homolog 3; Frizzled-3; Fz-3; Fzd3; hFz3; mFz3
Gene Aliases: AU020229; D930050A07Rik; Frizzled-3; Fz-3; Fz3; FZD3
UniProt ID: (Human) Q9NPG1, (Mouse) Q61086
Entrez Gene ID: (Horse) 100054732, (Pig) 100153078, (Human) 7976, (Dog) 486093, (Rabbit) 100339544, (Bovine) 445419, (Mouse) 14365, (Chinese hamster) 100751712, (Rat) 266715
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support