Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FZR1 Antibodies

          Invitrogen

          FZR1 Polyclonal Antibody

          View all (10) FZR1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite FZR1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          FZR1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FZR1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FZR1 Antibody (PA5-85303) in ICC/IF

          Immunocytochemistry analysis of FZR1 in HeLa cells fixed in 4% paraformaldehyde at RT for 15 min. Samples were incubated with FZR1 polyclonal antibody (Product # PA5-85303) using a dilution of 1:500 (green). Red: Phalloidin, a cytoskeleton marker, diluted at 1:100. Scale bar = 10 μm. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FZR1 Antibody in Immunocytochemistry (ICC/IF)
          FZR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FZR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FZR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FZR1 Antibody in Western Blot (WB)
          FZR1 Antibody in Western Blot (WB)
          FZR1 Antibody in Western Blot (WB)

          Product Details

          PA5-85303

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein encompassing a sequence within the center region of human FZR1. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.26 mg/mL

          Amount

          26 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 20% glycerol, 1% BSA

          Contains

          0.025% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2792446

          Product Specific Information

          Keep as concentrated solution.

          Predicted reactivity: Mouse (95%), Rat (95%), Xenopus laevis (90%), Pig (97%), Chicken (94%), Rhesus Monkey (100%), Bovine (97%).

          Positive Control: A431, A431 nuclear extract.

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          FZR is a 7WD domain family member implicated in cell cycle regulation of Drosophila and yeast. It has been shown that the over expression of fzr proteins increases natural killer cell-mediated cell death and as a result, suppresses tumor growth. The fzr protein is essential for cyclosome activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: APC regulator; CDC20 homolog 1; CDC20-like 1b; CDC20-like protein 1; CDH1; Cdh1/Hct1 homolog; cell division cycle 20 related 1; Fizzy-related protein homolog; fizzy/cell division cycle 20 related 1; Fzr; hCDH1

          View more View less

          Gene Aliases: AW108046; CDC20C; CDH1; DEE109; FYR; FZR; FZR1; FZR2; HCDH; HCDH1; KIAA1242

          View more View less

          UniProt ID: (Human) Q9UM11, (Mouse) Q9R1K5

          View more View less

          Entrez Gene ID: (Human) 51343, (Mouse) 56371, (Rat) 314642

          View more View less

          Function(s)
          protein binding anaphase-promoting complex binding ubiquitin-protein transferase activator activity protein binding, bridging involved in substrate recognition for ubiquitination ubiquitin ligase activator activity ubiquitin-protein ligase
          Process(es)
          DNA repair cellular response to DNA damage stimulus mitotic G2 DNA damage checkpoint regulation of mitotic cell cycle positive regulation of cell proliferation protein ubiquitination anaphase-promoting complex-dependent catabolic process regulation of meiotic nuclear division thymidine biosynthetic process cell division regulation of meiotic cell cycle proteolysis involved in cellular protein catabolic process lens fiber cell differentiation protein K11-linked ubiquitination positive regulation of ubiquitin protein ligase activity DNA synthesis involved in mitotic DNA replication positive regulation of anaphase-promoting complex-dependent catabolic process negative regulation of cellular senescence cell cycle mitotic nuclear division G2 DNA damage checkpoint positive regulation of protein catabolic process negative regulation of cell aging
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline