Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Fyn Antibodies

          Invitrogen

          Fyn Polyclonal Antibody

          Advanced Verification
          View all (35) Fyn antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Fyn Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          • Advanced Verification (1)
          Fyn Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Fyn Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Fyn Antibody (PA5-147912) in ICC/IF

          Immunofluorecent analysis of Fyn in human cell line AF22 using Fyn Polyclonal Antibody (Product # PA5-82851). Staining shows localization to plasma membrane. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Fyn Antibody in Immunocytochemistry (ICC/IF)
          Fyn Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Fyn Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Fyn Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Fyn Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Fyn Antibody in Western Blot (WB)
          Fyn Antibody in Western Blot (WB)
          Fyn Antibody
          Fyn Polyclonal Antibody

          Product Details

          PA5-147912

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.04-0.4 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          0.25-2 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein corresponding to Human FYN. Recombinant protein control fragment (Product #RP-95941).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.2, with 40% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          0

          Product Specific Information

          Immunogen sequence: SFGVTSIPNY NNFHAAGGQG LTVFGGVNSS SHTGTLRTRG GTGVTLFVAL YDYEARTEDD LSFHKGEKFQ ILNSSEGDWW EARSLTTGET GYIPSNYVAP VDSIQAEEWY FGKLGRKDAE RQLLSFGNPR GTFLIRESET TKGAYSLSIR

          Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

          Target Information

          FYN is a SRC-related kinase expressed in brain, skeletal muscle and adipose tissues. Fyn functions in a large number of pathways and has been shown to play a role in memory. FYN is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: C syn protooncogene; c-syn protooncogene; EC 2.7.10.2; FYN oncogene related to SRC, FGR, YES; fyn {ECO:0000312|RGD:2641}; kinase Fyn; MGC45350; OKT3-induced calcium influx regulator; OTTHUMP00000017916; OTTHUMP00000017917; P59 p59-Fyn; p59-Fyn; Proto-oncogene c-Fyn; Proto-oncogene Syn; proto-oncogene tyrosine-protein kinase Fyn; Protooncogene Syn; RP1-66H14.1; SLK; Src Kinase p59; Src like kinase; Src-like kinase; src-like tyrosine kinase (put.); putative; src/yes-related novel; tyrosine kinase p59fyn(T); Tyrosine-protein kinase Fyn; unnamed protein product

          View more View less

          Gene Aliases: AI448320; AW552119; FYN; p59-FYN; SLK; SYN

          View more View less

          UniProt ID: (Human) B5BU57, (Rat) Q62844, (Mouse) P39688

          View more View less

          Entrez Gene ID: (Human) 2534, (Rat) 25150, (Mouse) 14360

          View more View less

          Function(s)
          nucleotide binding G-protein coupled receptor binding protein kinase activity protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity receptor binding protein binding ATP binding tubulin binding phospholipase activator activity kinase activity transferase activity enzyme binding type 5 metabotropic glutamate receptor binding T cell receptor binding CD4 receptor binding CD8 receptor binding identical protein binding alpha-tubulin binding phospholipase binding phosphatidylinositol 3-kinase binding ion channel binding macromolecular complex binding metal ion binding ephrin receptor binding tau protein binding tau-protein kinase activity peptide hormone receptor binding growth factor receptor binding scaffold protein binding disordered domain specific binding glycoprotein binding protein complex binding transferase activity, transferring phosphorus-containing groups
          Process(es)
          response to singlet oxygen neuron migration stimulatory C-type lectin receptor signaling pathway adaptive immune response immune system process negative regulation of inflammatory response to antigenic stimulus heart process calcium ion transport cell surface receptor signaling pathway transmembrane receptor protein tyrosine kinase signaling pathway G-protein coupled glutamate receptor signaling pathway axon guidance learning feeding behavior regulation of cell shape response to xenobiotic stimulus gene expression negative regulation of gene expression negative regulation of hydrogen peroxide biosynthetic process positive regulation of neuron projection development negative regulation of angiogenesis protein ubiquitination peptidyl-tyrosine phosphorylation layer formation in cerebral cortex natural killer cell activation cell differentiation protein catabolic process forebrain development neuron projection development T cell costimulation negative regulation of protein ubiquitination intracellular signal transduction cellular response to platelet-derived growth factor stimulus reelin-mediated signaling pathway Fc-gamma receptor signaling pathway involved in phagocytosis negative regulation of protein catabolic process response to cocaine proteasome-mediated ubiquitin-dependent protein catabolic process negative regulation of neuron apoptotic process response to ethanol vascular endothelial growth factor receptor signaling pathway ephrin receptor signaling pathway dendrite morphogenesis activated T cell proliferation modulation of synaptic transmission T cell receptor signaling pathway leukocyte migration detection of mechanical stimulus involved in sensory perception of pain positive regulation of protein kinase B signaling cellular response to hydrogen peroxide cellular response to growth factor stimulus cellular response to peptide hormone stimulus cellular response to transforming growth factor beta stimulus positive regulation of protein targeting to membrane dendritic spine maintenance positive regulation of protein localization to nucleus regulation of glutamate receptor signaling pathway negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway negative regulation of dendritic spine maintenance response to amyloid-beta cellular response to beta-amyloid cellular response to L-glutamate cellular response to glycine positive regulation of protein localization to membrane regulation of calcium ion import across plasma membrane regulation of neuron migration negative regulation of extrinsic apoptotic signaling pathway in absence of ligand protein phosphorylation central nervous system development positive regulation of phosphatidylinositol 3-kinase signaling cell migration ionotropic glutamate receptor signaling pathway peptidyl-tyrosine autophosphorylation T cell activation regulation of cell proliferation response to drug positive regulation of I-kappaB kinase/NF-kappaB signaling innate immune response protein autophosphorylation multicellular organism development phosphorylation myelination regulation of apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline