Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LLSNSGQPE RKFMQTFVLA PEGSVPNKFY VHNDMFRYED EVFGDSEPEL DEESEDEVEE EQEERQPSPE PVQENANSGY YEAHPVTNGI EEPLEESSHE PEPEPESETK TEELKPQVEE KNLEELEEKS TTPPPAEPVS LPQEPPKPRV EAKPEVQSQP PRVREQRPRE RPGFPPRGPR PGRGDMEQND SDNRRIIRYP DSHQLFVGNL PHDIDENELK EFFMSFGNVV ELRINTKGVG GKLPNFGFVV FDDSEPVQRI LIAKPIMFRG EVRLNVEEKK TRAARERETR GGGDDRRDIR RNDRGPGGPR GIVGGGMMRD RDGRGPPPRG GMAQKLGSGR GTGQMEGRFT GQRR (97-449 aa encoded by BC011731)
G3BP2 gene ontology annotations related to this gene include stress granule assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alternatively spliced; G3BP-2; GAP SH3 domain-binding protein 2; GTPase activating protein (SH3 domain) binding protein 2; Ras-GTPase activating protein SH3 domain-binding protein 2; Rasputin homolog; unnamed protein product
UniProt ID: (Human) Q9UN86
Entrez Gene ID: (Human) 9908
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support